Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 135547071-135547322 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OBP2A
Entrez Gene ID
29991
Gene Name
odorant binding protein 2A
Gene Aliases
LCN13, OBP, OBP2C, OBPIIa, hOBPIIa
UniGene
Hs.567489
Gene Chromosome Location
Chr.9: 135544812-135549969 on Build GRCh38
Transcripts
NM_001293189.1 NM_001293193.1 NM_014582.2 NR_120603.1 XM_017014656.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1460979 c.245G>A p.R82Q Chr.9: 135547216-135547216 on Build GRCh38
5351581 c.256G>T p.E86* Chr.9: 135547227-135547227 on Build GRCh38
3655545 c.240G>A p.L80L Chr.9: 135547211-135547211 on Build GRCh38
3655543 c.226C>T p.Q76* Chr.9: 135547197-135547197 on Build GRCh38
5179341 c.245G>A p.R82Q Chr.9: 135547216-135547216 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs372804933 Chr.9: 135547223-135547223 on Build GRCh38
rs774865006 Chr.9: 135547188-135547188 on Build GRCh38
rs761834992 Chr.9: 135547133-135547134 on Build GRCh38
rs369099767 Chr.9: 135547189-135547189 on Build GRCh38
rs758273815 Chr.9: 135547164-135547164 on Build GRCh38
rs756618805 Chr.9: 135547281-135547281 on Build GRCh38
rs1047105902 Chr.9: 135547094-135547094 on Build GRCh38
rs757099879 Chr.9: 135547227-135547227 on Build GRCh38
rs768097168 Chr.9: 135547136-135547136 on Build GRCh38
rs778704196 Chr.9: 135547162-135547162 on Build GRCh38
rs376877476 Chr.9: 135547128-135547128 on Build GRCh38
rs762312791 Chr.9: 135547198-135547198 on Build GRCh38
rs756313978 Chr.9: 135547160-135547160 on Build GRCh38
rs750237306 Chr.9: 135547270-135547270 on Build GRCh38
rs200233464 Chr.9: 135547216-135547216 on Build GRCh38
rs751332323 Chr.9: 135547226-135547226 on Build GRCh38
rs201463203 Chr.9: 135547262-135547262 on Build GRCh38
rs111808171 Chr.9: 135547244-135547244 on Build GRCh38
rs776740042 Chr.9: 135547263-135547263 on Build GRCh38
rs757187744 Chr.9: 135547267-135547267 on Build GRCh38
rs759085810 Chr.9: 135547138-135547138 on Build GRCh38
rs492193 Chr.9: 135547240-135547240 on Build GRCh38
rs201539170 Chr.9: 135547154-135547154 on Build GRCh38
rs386739464 Chr.9: 135547134-135547136 on Build GRCh38
rs780814413 Chr.9: 135547231-135547231 on Build GRCh38
rs764448857 Chr.9: 135547156-135547156 on Build GRCh38
rs369119006 Chr.9: 135547246-135547246 on Build GRCh38
rs4006725 Chr.9: 135547136-135547136 on Build GRCh38
rs770592572 Chr.9: 135547135-135547135 on Build GRCh38
rs909988039 Chr.9: 135547095-135547095 on Build GRCh38
rs770539533 Chr.9: 135547173-135547173 on Build GRCh38
rs750930487 Chr.9: 135547274-135547274 on Build GRCh38
rs751563569 Chr.9: 135547265-135547265 on Build GRCh38
rs892891945 Chr.9: 135547294-135547294 on Build GRCh38
rs370379380 Chr.9: 135547127-135547127 on Build GRCh38
rs573532618 Chr.9: 135547108-135547108 on Build GRCh38
rs746837596 Chr.9: 135547167-135547167 on Build GRCh38
rs779562489 Chr.9: 135547291-135547291 on Build GRCh38
rs368244666 Chr.9: 135547245-135547245 on Build GRCh38
rs769357569 Chr.9: 135547181-135547181 on Build GRCh38
rs504061 Chr.9: 135547159-135547159 on Build GRCh38
rs367856133 Chr.9: 135547131-135547131 on Build GRCh38
rs751654193 Chr.9: 135547134-135547150 on Build GRCh38
rs532869593 Chr.9: 135547125-135547125 on Build GRCh38
rs78161279 Chr.9: 135547130-135547130 on Build GRCh38
rs566194944 Chr.9: 135547180-135547180 on Build GRCh38
rs1007745433 Chr.9: 135547300-135547300 on Build GRCh38
rs373739564 Chr.9: 135547264-135547264 on Build GRCh38
rs748600666 Chr.9: 135547292-135547292 on Build GRCh38
rs12344721 Chr.9: 135547269-135547269 on Build GRCh38
rs535886153 Chr.9: 135547158-135547158 on Build GRCh38
rs777567300 Chr.9: 135547166-135547166 on Build GRCh38
rs774741757 Chr.9: 135547190-135547190 on Build GRCh38
rs780869544 Chr.9: 135547177-135547177 on Build GRCh38
rs559436873 Chr.9: 135547120-135547120 on Build GRCh38
rs200668491 Chr.9: 135547289-135547289 on Build GRCh38
rs374742179 Chr.9: 135547290-135547290 on Build GRCh38
rs750734814 Chr.9: 135547213-135547213 on Build GRCh38
rs4006724 Chr.9: 135547134-135547134 on Build GRCh38
rs772632803 Chr.9: 135547249-135547249 on Build GRCh38
rs745455326 Chr.9: 135547235-135547235 on Build GRCh38
rs373559473 Chr.9: 135547148-135547148 on Build GRCh38
rs376053905 Chr.9: 135547260-135547260 on Build GRCh38
rs139660402 Chr.9: 135547215-135547215 on Build GRCh38
rs540666833 Chr.9: 135547111-135547111 on Build GRCh38
rs767832694 Chr.9: 135547200-135547200 on Build GRCh38
rs773718900 Chr.9: 135547259-135547259 on Build GRCh38
rs200528184 Chr.9: 135547151-135547151 on Build GRCh38
rs149715359 Chr.9: 135547222-135547222 on Build GRCh38

View Full Product Details