Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 24918337-24918578 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NEFM
Entrez Gene ID
4741
Gene Name
neurofilament, medium polypeptide
Gene Aliases
NEF3, NF-M, NFM
UniGene
Hs.458657
Gene Chromosome Location
Chr.8: 24913761-24919093 on Build GRCh38
Transcripts
NM_001105541.1 NM_005382.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5450466 c.2661A>C p.E887D Chr.8: 24918516-24918516 on Build GRCh38
5450465 c.2634C>T p.T878T Chr.8: 24918489-24918489 on Build GRCh38
4706369 c.2718C>T p.H906H Chr.8: 24918573-24918573 on Build GRCh38
1258955 c.2552A>C p.K851T Chr.8: 24918407-24918407 on Build GRCh38
4706368 c.2528_2529insG p.D846fs*1 Chr.8: 24918383-24918384 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774135441 Chr.8: 24918410-24918410 on Build GRCh38
rs865855175 Chr.8: 24918376-24918376 on Build GRCh38
rs749966909 Chr.8: 24918392-24918392 on Build GRCh38
rs779188160 Chr.8: 24918374-24918374 on Build GRCh38
rs373997729 Chr.8: 24918425-24918425 on Build GRCh38
rs57658073 Chr.8: 24918427-24918427 on Build GRCh38
rs750609179 Chr.8: 24918443-24918443 on Build GRCh38
rs756498993 Chr.8: 24918476-24918476 on Build GRCh38
rs765423550 Chr.8: 24918436-24918436 on Build GRCh38
rs756479994 Chr.8: 24918467-24918467 on Build GRCh38
rs144200733 Chr.8: 24918449-24918449 on Build GRCh38
rs765108284 Chr.8: 24918516-24918516 on Build GRCh38
rs753638891 Chr.8: 24918426-24918426 on Build GRCh38
rs758582875 Chr.8: 24918448-24918448 on Build GRCh38
rs763178564 Chr.8: 24918546-24918546 on Build GRCh38
rs770556393 Chr.8: 24918402-24918402 on Build GRCh38
rs547142877 Chr.8: 24918355-24918355 on Build GRCh38
rs775088662 Chr.8: 24918415-24918415 on Build GRCh38
rs140175409 Chr.8: 24918452-24918452 on Build GRCh38
rs111429784 Chr.8: 24918530-24918530 on Build GRCh38
rs750555881 Chr.8: 24918383-24918383 on Build GRCh38
rs200419471 Chr.8: 24918474-24918474 on Build GRCh38
rs748520435 Chr.8: 24918460-24918460 on Build GRCh38
rs140995374 Chr.8: 24918490-24918490 on Build GRCh38
rs758449006 Chr.8: 24918388-24918388 on Build GRCh38
rs770245529 Chr.8: 24918534-24918534 on Build GRCh38
rs199870416 Chr.8: 24918478-24918478 on Build GRCh38
rs771737027 Chr.8: 24918414-24918414 on Build GRCh38
rs147964775 Chr.8: 24918450-24918450 on Build GRCh38
rs151082365 Chr.8: 24918489-24918489 on Build GRCh38
rs781720571 Chr.8: 24918455-24918455 on Build GRCh38
rs185587224 Chr.8: 24918503-24918503 on Build GRCh38
rs944891517 Chr.8: 24918521-24918521 on Build GRCh38
rs753611085 Chr.8: 24918363-24918363 on Build GRCh38
rs778201930 Chr.8: 24918510-24918510 on Build GRCh38
rs745535854 Chr.8: 24918477-24918477 on Build GRCh38
rs188421098 Chr.8: 24918389-24918389 on Build GRCh38
rs757425078 Chr.8: 24918372-24918372 on Build GRCh38
rs150229714 Chr.8: 24918508-24918508 on Build GRCh38
rs143878463 Chr.8: 24918390-24918390 on Build GRCh38
rs759167285 Chr.8: 24918412-24918412 on Build GRCh38
rs369756657 Chr.8: 24918444-24918444 on Build GRCh38
rs781427544 Chr.8: 24918391-24918391 on Build GRCh38

View Full Product Details