Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 34122390-34122651 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NAT10
Entrez Gene ID
55226
Gene Name
N-acetyltransferase 10
Gene Aliases
ALP, NET43
UniGene
Hs.577281
Gene Chromosome Location
Chr.11: 34105564-34146911 on Build GRCh38
Transcripts
NM_001144030.1 NM_024662.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4032511 c.806A>T p.E269V Chr.11: 34122484-34122484 on Build GRCh38
926605 c.908C>T p.A303V Chr.11: 34122586-34122586 on Build GRCh38
926604 c.822G>T p.K274N Chr.11: 34122500-34122500 on Build GRCh38
5886733 c.830G>A p.R277K Chr.11: 34122508-34122508 on Build GRCh38
6315434 c.790G>T p.V264F Chr.11: 34122468-34122468 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746779535 Chr.11: 34122587-34122587 on Build GRCh38
rs749425253 Chr.11: 34122446-34122447 on Build GRCh38
rs1035595686 Chr.11: 34122457-34122457 on Build GRCh38
rs201938195 Chr.11: 34122418-34122418 on Build GRCh38
rs768552847 Chr.11: 34122595-34122595 on Build GRCh38
rs112208767 Chr.11: 34122630-34122630 on Build GRCh38
rs748712610 Chr.11: 34122487-34122487 on Build GRCh38
rs759270288 Chr.11: 34122505-34122505 on Build GRCh38
rs775322184 Chr.11: 34122582-34122582 on Build GRCh38
rs370205958 Chr.11: 34122568-34122568 on Build GRCh38
rs201610017 Chr.11: 34122580-34122580 on Build GRCh38
rs113578380 Chr.11: 34122515-34122515 on Build GRCh38
rs1052004026 Chr.11: 34122483-34122483 on Build GRCh38
rs749876874 Chr.11: 34122560-34122560 on Build GRCh38
rs114351327 Chr.11: 34122453-34122453 on Build GRCh38
rs939710595 Chr.11: 34122531-34122531 on Build GRCh38
rs751865245 Chr.11: 34122450-34122450 on Build GRCh38
rs759711601 Chr.11: 34122616-34122616 on Build GRCh38
rs955997925 Chr.11: 34122598-34122598 on Build GRCh38
rs776678760 Chr.11: 34122516-34122516 on Build GRCh38
rs766545139 Chr.11: 34122540-34122540 on Build GRCh38
rs146301468 Chr.11: 34122542-34122542 on Build GRCh38
rs758546728 Chr.11: 34122441-34122441 on Build GRCh38
rs142807420 Chr.11: 34122581-34122581 on Build GRCh38
rs754416795 Chr.11: 34122535-34122535 on Build GRCh38
rs373535671 Chr.11: 34122541-34122541 on Build GRCh38
rs376383993 Chr.11: 34122559-34122559 on Build GRCh38
rs776428666 Chr.11: 34122608-34122608 on Build GRCh38
rs761813028 Chr.11: 34122609-34122609 on Build GRCh38
rs771936992 Chr.11: 34122506-34122506 on Build GRCh38
rs142291579 Chr.11: 34122482-34122482 on Build GRCh38
rs767690927 Chr.11: 34122622-34122622 on Build GRCh38
rs773872950 Chr.11: 34122488-34122488 on Build GRCh38
rs975274958 Chr.11: 34122452-34122452 on Build GRCh38
rs376848619 Chr.11: 34122466-34122466 on Build GRCh38
rs760650469 Chr.11: 34122514-34122514 on Build GRCh38
rs1010631947 Chr.11: 34122600-34122600 on Build GRCh38
rs371140580 Chr.11: 34122480-34122480 on Build GRCh38
rs762927840 Chr.11: 34122524-34122524 on Build GRCh38
rs139466580 Chr.11: 34122569-34122569 on Build GRCh38
rs912210828 Chr.11: 34122513-34122513 on Build GRCh38
rs758573486 Chr.11: 34122520-34122520 on Build GRCh38
rs766310851 Chr.11: 34122613-34122613 on Build GRCh38
rs376882338 Chr.11: 34122432-34122432 on Build GRCh38
rs140175046 Chr.11: 34122468-34122468 on Build GRCh38
rs376218350 Chr.11: 34122614-34122614 on Build GRCh38
rs151136899 Chr.11: 34122485-34122485 on Build GRCh38
rs369179275 Chr.11: 34122440-34122440 on Build GRCh38
rs775450573 Chr.11: 34122508-34122508 on Build GRCh38
rs780116927 Chr.11: 34122442-34122442 on Build GRCh38
rs756507373 Chr.11: 34122555-34122555 on Build GRCh38

View Full Product Details