Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 63248600-63248873 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NKAIN4
Entrez Gene ID
128414
Gene Name
Na+/K+ transporting ATPase interacting 4
Gene Aliases
C20orf58, FAM77A, bA261N11.2
UniGene
Hs.46627
Gene Chromosome Location
Chr.20: 63240784-63254904 on Build GRCh38
Transcripts
NM_152864.3 XM_005260192.1 XM_005260194.3 XM_011528527.1 XM_011528528.2 XM_011528529.2 XM_017027636.1 XM_017027637.1 XM_017027638.1 XM_017027639.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7300337 c.263G>A p.G88D Chr.20: 63248825-63248825 on Build GRCh38
4400487 c.265C>T p.L89F Chr.20: 63248823-63248823 on Build GRCh38
4939767 c.227A>G p.N76S Chr.20: 63248861-63248861 on Build GRCh38
5031448 c.249C>A p.Y83* Chr.20: 63248839-63248839 on Build GRCh38
6353420 c.271A>C p.K91Q Chr.20: 63248817-63248817 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761300876 Chr.20: 63248803-63248803 on Build GRCh38
rs117617608 Chr.20: 63248791-63248791 on Build GRCh38
rs772566328 Chr.20: 63248784-63248784 on Build GRCh38
rs6090331 Chr.20: 63248675-63248675 on Build GRCh38
rs771333438 Chr.20: 63248765-63248765 on Build GRCh38
rs1129659 Chr.20: 63248817-63248817 on Build GRCh38
rs980205277 Chr.20: 63248689-63248689 on Build GRCh38
rs963581904 Chr.20: 63248671-63248671 on Build GRCh38
rs894301813 Chr.20: 63248701-63248701 on Build GRCh38
rs578211100 Chr.20: 63248749-63248749 on Build GRCh38
rs537047390 Chr.20: 63248724-63248724 on Build GRCh38
rs375389370 Chr.20: 63248767-63248767 on Build GRCh38
rs950608482 Chr.20: 63248645-63248645 on Build GRCh38
rs1005880014 Chr.20: 63248685-63248685 on Build GRCh38
rs7261433 Chr.20: 63248760-63248760 on Build GRCh38
rs913263334 Chr.20: 63248730-63248730 on Build GRCh38
rs556233671 Chr.20: 63248810-63248810 on Build GRCh38
rs922701691 Chr.20: 63248814-63248814 on Build GRCh38
rs749678776 Chr.20: 63248723-63248723 on Build GRCh38
rs757397579 Chr.20: 63248816-63248816 on Build GRCh38
rs78017705 Chr.20: 63248721-63248721 on Build GRCh38
rs190913394 Chr.20: 63248792-63248792 on Build GRCh38
rs930012294 Chr.20: 63248637-63248637 on Build GRCh38
rs760151334 Chr.20: 63248772-63248772 on Build GRCh38
rs138254796 Chr.20: 63248824-63248824 on Build GRCh38
rs536402112 Chr.20: 63248656-63248656 on Build GRCh38
rs926132775 Chr.20: 63248761-63248761 on Build GRCh38
rs553121179 Chr.20: 63248697-63248697 on Build GRCh38
rs867150672 Chr.20: 63248835-63248835 on Build GRCh38
rs770442104 Chr.20: 63248726-63248726 on Build GRCh38
rs141264539 Chr.20: 63248853-63248853 on Build GRCh38
rs749538983 Chr.20: 63248847-63248847 on Build GRCh38
rs766673619 Chr.20: 63248657-63248657 on Build GRCh38
rs753955657 Chr.20: 63248708-63248708 on Build GRCh38
rs749433186 Chr.20: 63248764-63248764 on Build GRCh38
rs556789198 Chr.20: 63248748-63248748 on Build GRCh38
rs565172154 Chr.20: 63248829-63248829 on Build GRCh38
rs6122411 Chr.20: 63248795-63248795 on Build GRCh38
rs781105496 Chr.20: 63248853-63248860 on Build GRCh38
rs199879209 Chr.20: 63248790-63248790 on Build GRCh38
rs35537426 Chr.20: 63248830-63248830 on Build GRCh38
rs201793448 Chr.20: 63248823-63248823 on Build GRCh38
rs142657484 Chr.20: 63248826-63248826 on Build GRCh38
rs976864595 Chr.20: 63248799-63248799 on Build GRCh38
rs375719568 Chr.20: 63248787-63248787 on Build GRCh38

View Full Product Details