Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 40745164-40745389 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RMDN3
Entrez Gene ID
55177
Gene Name
regulator of microtubule dynamics 3
Gene Aliases
FAM82A2, FAM82C, RMD-3, RMD3, ptpip51
UniGene
Hs.511067
Gene Chromosome Location
Chr.15: 40735884-40755336 on Build GRCh38
Transcripts
NM_001304802.1 NM_001323894.1 NM_001323895.1 NM_001323896.1 NM_001323897.1 NM_018145.2 XM_011521755.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4441638 c.560G>A p.R187Q Chr.15: 40745224-40745224 on Build GRCh38
1301080 c.562G>A p.D188N Chr.15: 40745222-40745222 on Build GRCh38
3815981 c.556G>A p.E186K Chr.15: 40745228-40745228 on Build GRCh38
6767034 c.561G>A p.R187R Chr.15: 40745223-40745223 on Build GRCh38
160973 c.525-1G>C p.? Chr.15: 40745260-40745260 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs34847558 Chr.15: 40745313-40745313 on Build GRCh38
rs369389009 Chr.15: 40745209-40745209 on Build GRCh38
rs926888248 Chr.15: 40745334-40745334 on Build GRCh38
rs767117089 Chr.15: 40745304-40745304 on Build GRCh38
rs779057197 Chr.15: 40745229-40745229 on Build GRCh38
rs768144583 Chr.15: 40745206-40745209 on Build GRCh38
rs752514679 Chr.15: 40745326-40745326 on Build GRCh38
rs767256940 Chr.15: 40745241-40745241 on Build GRCh38
rs755989202 Chr.15: 40745207-40745207 on Build GRCh38
rs35952852 Chr.15: 40745280-40745280 on Build GRCh38
rs762671209 Chr.15: 40745191-40745191 on Build GRCh38
rs981009125 Chr.15: 40745331-40745331 on Build GRCh38
rs183230311 Chr.15: 40745212-40745212 on Build GRCh38
rs774190360 Chr.15: 40745290-40745290 on Build GRCh38
rs762220979 Chr.15: 40745287-40745287 on Build GRCh38
rs780996472 Chr.15: 40745280-40745282 on Build GRCh38
rs114290296 Chr.15: 40745284-40745284 on Build GRCh38
rs766073179 Chr.15: 40745193-40745193 on Build GRCh38
rs78037363 Chr.15: 40745319-40745319 on Build GRCh38
rs754502218 Chr.15: 40745198-40745198 on Build GRCh38
rs910097337 Chr.15: 40745350-40745350 on Build GRCh38
rs1028895848 Chr.15: 40745186-40745186 on Build GRCh38
rs377576031 Chr.15: 40745199-40745199 on Build GRCh38
rs372835028 Chr.15: 40745222-40745222 on Build GRCh38
rs761686647 Chr.15: 40745204-40745204 on Build GRCh38
rs771152595 Chr.15: 40745225-40745225 on Build GRCh38
rs936917302 Chr.15: 40745335-40745335 on Build GRCh38
rs766163395 Chr.15: 40745289-40745289 on Build GRCh38
rs772178902 Chr.15: 40745237-40745237 on Build GRCh38
rs76383777 Chr.15: 40745318-40745318 on Build GRCh38
rs759245973 Chr.15: 40745299-40745299 on Build GRCh38
rs751140051 Chr.15: 40745195-40745195 on Build GRCh38
rs144106216 Chr.15: 40745242-40745242 on Build GRCh38
rs752732958 Chr.15: 40745202-40745202 on Build GRCh38
rs960564928 Chr.15: 40745245-40745245 on Build GRCh38
rs377060294 Chr.15: 40745232-40745232 on Build GRCh38
rs1048737733 Chr.15: 40745308-40745308 on Build GRCh38
rs207475432 Chr.15: 40745268-40745268 on Build GRCh38
rs536641013 Chr.15: 40745316-40745316 on Build GRCh38
rs752139998 Chr.15: 40745305-40745305 on Build GRCh38
rs748966326 Chr.15: 40745224-40745224 on Build GRCh38

View Full Product Details