Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 45420223-45420448 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PIGT
Entrez Gene ID
51604
Gene Name
phosphatidylinositol glycan anchor biosynthesis class T
Gene Aliases
CGI-06, MCAHS3, NDAP, PNH2
UniGene
Hs.437388
Gene Chromosome Location
Chr.20: 45416067-45426245 on Build GRCh38
Transcripts
NM_001184728.2 NM_001184729.2 NM_001184730.2 NM_015937.5 NR_047691.1 NR_047692.1 NR_047693.1 NR_047694.1 NR_047695.1 XM_005260430.2 XM_005260432.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7322492 c.769G>T p.D257Y Chr.20: 45420223-45420223 on Build GRCh38
3939368 c.825T>C p.A275A Chr.20: 45420387-45420387 on Build GRCh38
135882 c.818C>T p.P273L Chr.20: 45420380-45420380 on Build GRCh38
7383900 c.812C>T p.P271L Chr.20: 45420374-45420374 on Build GRCh38
3547023 c.792C>T p.F264F Chr.20: 45420354-45420354 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762698156 Chr.20: 45420284-45420285 on Build GRCh38
rs751236460 Chr.20: 45420409-45420409 on Build GRCh38
rs202220796 Chr.20: 45420398-45420398 on Build GRCh38
rs372245517 Chr.20: 45420318-45420318 on Build GRCh38
rs754145899 Chr.20: 45420346-45420346 on Build GRCh38
rs764520337 Chr.20: 45420339-45420339 on Build GRCh38
rs768633876 Chr.20: 45420274-45420274 on Build GRCh38
rs368919212 Chr.20: 45420323-45420323 on Build GRCh38
rs574857016 Chr.20: 45420410-45420410 on Build GRCh38
rs756126000 Chr.20: 45420428-45420428 on Build GRCh38
rs746565696 Chr.20: 45420356-45420356 on Build GRCh38
rs762138208 Chr.20: 45420401-45420401 on Build GRCh38
rs779662920 Chr.20: 45420348-45420348 on Build GRCh38
rs199968454 Chr.20: 45420397-45420397 on Build GRCh38
rs149740779 Chr.20: 45420368-45420368 on Build GRCh38
rs774743095 Chr.20: 45420374-45420374 on Build GRCh38
rs759782266 Chr.20: 45420380-45420380 on Build GRCh38
rs368372554 Chr.20: 45420307-45420307 on Build GRCh38
rs746512712 Chr.20: 45420269-45420269 on Build GRCh38
rs759656534 Chr.20: 45420319-45420319 on Build GRCh38
rs766694784 Chr.20: 45420317-45420317 on Build GRCh38
rs776521852 Chr.20: 45420281-45420281 on Build GRCh38
rs544841186 Chr.20: 45420404-45420404 on Build GRCh38
rs779602919 Chr.20: 45420258-45420258 on Build GRCh38
rs771317926 Chr.20: 45420369-45420369 on Build GRCh38
rs368747168 Chr.20: 45420367-45420367 on Build GRCh38
rs139713205 Chr.20: 45420363-45420363 on Build GRCh38
rs993697819 Chr.20: 45420381-45420381 on Build GRCh38
rs768495541 Chr.20: 45420367-45420368 on Build GRCh38
rs775362454 Chr.20: 45420392-45420392 on Build GRCh38
rs780632511 Chr.20: 45420359-45420359 on Build GRCh38
rs191780806 Chr.20: 45420306-45420306 on Build GRCh38
rs989674587 Chr.20: 45420280-45420280 on Build GRCh38
rs140324917 Chr.20: 45420411-45420411 on Build GRCh38
rs945509622 Chr.20: 45420292-45420292 on Build GRCh38
rs370743585 Chr.20: 45420302-45420302 on Build GRCh38
rs574145282 Chr.20: 45420288-45420288 on Build GRCh38
rs767662825 Chr.20: 45420331-45420331 on Build GRCh38
rs544502973 Chr.20: 45420289-45420289 on Build GRCh38
rs757918086 Chr.20: 45420251-45420251 on Build GRCh38
rs767715699 Chr.20: 45420387-45420387 on Build GRCh38
rs35320070 Chr.20: 45420426-45420426 on Build GRCh38
rs146764121 Chr.20: 45420399-45420399 on Build GRCh38
rs61753669 Chr.20: 45420332-45420332 on Build GRCh38
rs578178905 Chr.20: 45420347-45420347 on Build GRCh38
rs374597406 Chr.20: 45420320-45420320 on Build GRCh38
rs754517456 Chr.20: 45420358-45420358 on Build GRCh38
rs756518985 Chr.20: 45420338-45420338 on Build GRCh38

View Full Product Details