Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 38401048-38401307 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OTC
Entrez Gene ID
5009
Gene Name
ornithine carbamoyltransferase
Gene Aliases
OCTD
UniGene
Hs.117050
Gene Chromosome Location
Chr.X: 38352483-38421450 on Build GRCh38
Transcripts
NM_000531.5 XM_017029556.1

SNP Detail

SNP ID Chromosome Location
rs928860938 Chr.X: 38401175-38401175 on Build GRCh38
rs72556254 Chr.X: 38401283-38401283 on Build GRCh38
rs368240058 Chr.X: 38401226-38401226 on Build GRCh38
rs72556251 Chr.X: 38401278-38401279 on Build GRCh38
rs773392694 Chr.X: 38401139-38401139 on Build GRCh38
rs375307125 Chr.X: 38401250-38401250 on Build GRCh38
rs747580006 Chr.X: 38401239-38401239 on Build GRCh38
rs180977607 Chr.X: 38401074-38401074 on Build GRCh38
rs149795059 Chr.X: 38401278-38401278 on Build GRCh38
rs769794826 Chr.X: 38401081-38401081 on Build GRCh38
rs66556380 Chr.X: 38401273-38401273 on Build GRCh38
rs1052206430 Chr.X: 38401275-38401275 on Build GRCh38
rs748353850 Chr.X: 38401247-38401247 on Build GRCh38
rs773527760 Chr.X: 38401251-38401251 on Build GRCh38
rs758845954 Chr.X: 38401224-38401224 on Build GRCh38
rs947804300 Chr.X: 38401143-38401143 on Build GRCh38
rs72556252 Chr.X: 38401280-38401280 on Build GRCh38
rs1046180504 Chr.X: 38401221-38401221 on Build GRCh38
rs1800326 Chr.X: 38401268-38401268 on Build GRCh38
rs183600147 Chr.X: 38401213-38401213 on Build GRCh38
rs746896364 Chr.X: 38401080-38401080 on Build GRCh38
rs370882565 Chr.X: 38401243-38401243 on Build GRCh38
rs72556253 Chr.X: 38401282-38401282 on Build GRCh38

View Full Product Details