Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 149023964-149024189 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GYG1
Entrez Gene ID
2992
Gene Name
glycogenin 1
Gene Aliases
GSD15, GYG
UniGene
Hs.477892
Gene Chromosome Location
Chr.3: 148991408-149027669 on Build GRCh38
Transcripts
NM_001184720.1 NM_001184721.1 NM_004130.3 XM_017006275.1 XM_017006276.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3427193 c.736C>A p.L246I Chr.3: 149024180-149024180 on Build GRCh38
1419818 c.684_685delAA p.S230fs*4 Chr.3: 149024128-149024129 on Build GRCh38
6011235 c.665A>T p.Y222F Chr.3: 149024109-149024109 on Build GRCh38
4114326 c.717C>T p.N239N Chr.3: 149024161-149024161 on Build GRCh38
3002563 c.710A>C p.D237A Chr.3: 149024154-149024154 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1009324457 Chr.3: 149024115-149024115 on Build GRCh38
rs780894437 Chr.3: 149024104-149024105 on Build GRCh38
rs748162789 Chr.3: 149024034-149024034 on Build GRCh38
rs751953419 Chr.3: 149024070-149024070 on Build GRCh38
rs75445811 Chr.3: 149024053-149024053 on Build GRCh38
rs759134229 Chr.3: 149024160-149024160 on Build GRCh38
rs750552953 Chr.3: 149024057-149024057 on Build GRCh38
rs998673604 Chr.3: 149024037-149024037 on Build GRCh38
rs1004038883 Chr.3: 149024078-149024078 on Build GRCh38
rs368838737 Chr.3: 149024033-149024033 on Build GRCh38
rs754678008 Chr.3: 149024021-149024024 on Build GRCh38
rs1035133767 Chr.3: 149024091-149024091 on Build GRCh38
rs768737015 Chr.3: 149024040-149024040 on Build GRCh38
rs146085961 Chr.3: 149024042-149024042 on Build GRCh38
rs749552019 Chr.3: 149024129-149024129 on Build GRCh38
rs771963932 Chr.3: 149024118-149024118 on Build GRCh38
rs768562883 Chr.3: 149024145-149024145 on Build GRCh38
rs756870155 Chr.3: 149024100-149024100 on Build GRCh38
rs375493988 Chr.3: 149024006-149024006 on Build GRCh38
rs560773197 Chr.3: 149024037-149024038 on Build GRCh38
rs747627650 Chr.3: 149024127-149024127 on Build GRCh38
rs201672568 Chr.3: 149024127-149024127 on Build GRCh38
rs769385760 Chr.3: 149024128-149024128 on Build GRCh38
rs920742617 Chr.3: 149024151-149024151 on Build GRCh38
rs780014472 Chr.3: 149024124-149024124 on Build GRCh38
rs372805607 Chr.3: 149024144-149024144 on Build GRCh38
rs532571990 Chr.3: 149024114-149024114 on Build GRCh38
rs776294715 Chr.3: 149024149-149024149 on Build GRCh38
rs567161281 Chr.3: 149024054-149024054 on Build GRCh38
rs755431625 Chr.3: 149024077-149024077 on Build GRCh38
rs753467119 Chr.3: 149024094-149024094 on Build GRCh38
rs956327847 Chr.3: 149024101-149024101 on Build GRCh38
rs763516254 Chr.3: 149024058-149024058 on Build GRCh38
rs777291159 Chr.3: 149024090-149024090 on Build GRCh38
rs762046425 Chr.3: 149024150-149024150 on Build GRCh38
rs199593097 Chr.3: 149024039-149024039 on Build GRCh38
rs767034971 Chr.3: 149024064-149024064 on Build GRCh38
rs745411888 Chr.3: 149024116-149024116 on Build GRCh38

View Full Product Details