Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 117210007-117210232 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNK18
Entrez Gene ID
338567
Gene Name
potassium two pore domain channel subfamily K member 18
Gene Aliases
K2p18.1, MGR13, TRESK, TRESK-2, TRESK2, TRIK
UniGene
Hs.449650
Gene Chromosome Location
Chr.10: 117197489-117210299 on Build GRCh38
Transcripts
NM_181840.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2055033 c.863T>A p.L288H Chr.10: 117210007-117210007 on Build GRCh38
5205794 c.869delT p.F291fs*41 Chr.10: 117210013-117210013 on Build GRCh38
6537130 c.970A>G p.I324V Chr.10: 117210114-117210114 on Build GRCh38
2055037 c.916_917delAC p.Q307fs*28 Chr.10: 117210060-117210061 on Build GRCh38
5037827 c.1081A>C p.R361R Chr.10: 117210225-117210225 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771838477 Chr.10: 117210094-117210094 on Build GRCh38
rs368800855 Chr.10: 117210115-117210115 on Build GRCh38
rs770869540 Chr.10: 117210175-117210175 on Build GRCh38
rs147749657 Chr.10: 117210151-117210151 on Build GRCh38
rs982208755 Chr.10: 117210050-117210050 on Build GRCh38
rs764660644 Chr.10: 117210187-117210187 on Build GRCh38
rs145335768 Chr.10: 117210066-117210066 on Build GRCh38
rs745940715 Chr.10: 117210145-117210145 on Build GRCh38
rs376558803 Chr.10: 117210110-117210110 on Build GRCh38
rs752379870 Chr.10: 117210197-117210197 on Build GRCh38
rs760575756 Chr.10: 117210174-117210174 on Build GRCh38
rs558023580 Chr.10: 117210155-117210155 on Build GRCh38
rs753117927 Chr.10: 117210119-117210119 on Build GRCh38
rs753322636 Chr.10: 117210112-117210112 on Build GRCh38
rs757761673 Chr.10: 117210141-117210141 on Build GRCh38
rs773024059 Chr.10: 117210109-117210109 on Build GRCh38
rs373933439 Chr.10: 117210074-117210074 on Build GRCh38
rs1034661804 Chr.10: 117210068-117210068 on Build GRCh38
rs771791105 Chr.10: 117210161-117210161 on Build GRCh38
rs758767008 Chr.10: 117210123-117210123 on Build GRCh38
rs954177406 Chr.10: 117210082-117210082 on Build GRCh38
rs770948780 Chr.10: 117210059-117210059 on Build GRCh38
rs374314144 Chr.10: 117210179-117210179 on Build GRCh38
rs759847537 Chr.10: 117210149-117210151 on Build GRCh38
rs773008291 Chr.10: 117210168-117210168 on Build GRCh38
rs778160917 Chr.10: 117210128-117210128 on Build GRCh38
rs774358745 Chr.10: 117210075-117210075 on Build GRCh38
rs752031342 Chr.10: 117210134-117210134 on Build GRCh38
rs12247136 Chr.10: 117210180-117210180 on Build GRCh38
rs747483367 Chr.10: 117210045-117210045 on Build GRCh38
rs764864752 Chr.10: 117210116-117210116 on Build GRCh38
rs780440795 Chr.10: 117210153-117210153 on Build GRCh38
rs563802272 Chr.10: 117210113-117210113 on Build GRCh38
rs548343681 Chr.10: 117210143-117210143 on Build GRCh38
rs528686489 Chr.10: 117210080-117210080 on Build GRCh38

View Full Product Details