Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 70132916-70133171 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNJ16
Entrez Gene ID
3773
Gene Name
potassium voltage-gated channel subfamily J member 16
Gene Aliases
BIR9, KIR5.1
UniGene
Hs.463985
Gene Chromosome Location
Chr.17: 70075225-70135608 on Build GRCh38
Transcripts
NM_001270422.1 NM_001291622.1 NM_001291623.1 NM_001291624.1 NM_001291625.1 NM_018658.2 NM_170741.2 NM_170742.2 XM_005257337.4 XM_006721885.3 XM_006721886.3 XM_006721887.3 XM_011524781.2 XM_017024609.1 XM_017024610.1 XM_017024611.1 XM_017024612.1 XM_017024613.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
983649 c.1042G>A p.D348N Chr.17: 70133129-70133129 on Build GRCh38
332719 c.903C>A p.P301P Chr.17: 70132990-70132990 on Build GRCh38
983648 c.907G>T p.E303* Chr.17: 70132994-70132994 on Build GRCh38
4069095 c.899T>A p.V300D Chr.17: 70132986-70132986 on Build GRCh38
5578783 c.1075G>A p.V359I Chr.17: 70133162-70133162 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772113222 Chr.17: 70133032-70133032 on Build GRCh38
rs747923234 Chr.17: 70133014-70133014 on Build GRCh38
rs749293591 Chr.17: 70133027-70133027 on Build GRCh38
rs751133691 Chr.17: 70133112-70133112 on Build GRCh38
rs763595340 Chr.17: 70133103-70133103 on Build GRCh38
rs768794203 Chr.17: 70133070-70133070 on Build GRCh38
rs148035314 Chr.17: 70132960-70132960 on Build GRCh38
rs773849972 Chr.17: 70133102-70133102 on Build GRCh38
rs754176962 Chr.17: 70132951-70132951 on Build GRCh38
rs967210348 Chr.17: 70133059-70133059 on Build GRCh38
rs986208220 Chr.17: 70133140-70133140 on Build GRCh38
rs765753898 Chr.17: 70133097-70133097 on Build GRCh38
rs765669108 Chr.17: 70132975-70132975 on Build GRCh38
rs777676628 Chr.17: 70133024-70133024 on Build GRCh38
rs747205925 Chr.17: 70133050-70133050 on Build GRCh38
rs908055201 Chr.17: 70133084-70133084 on Build GRCh38
rs146035669 Chr.17: 70132992-70132992 on Build GRCh38
rs953010349 Chr.17: 70133135-70133135 on Build GRCh38
rs776989799 Chr.17: 70133079-70133079 on Build GRCh38
rs754617006 Chr.17: 70133130-70133130 on Build GRCh38
rs138191197 Chr.17: 70133006-70133006 on Build GRCh38
rs757966954 Chr.17: 70132983-70132983 on Build GRCh38
rs765229418 Chr.17: 70133026-70133026 on Build GRCh38
rs780987537 Chr.17: 70133008-70133008 on Build GRCh38
rs35893652 Chr.17: 70133026-70133027 on Build GRCh38
rs201278174 Chr.17: 70132976-70132976 on Build GRCh38
rs769703462 Chr.17: 70133019-70133019 on Build GRCh38
rs775596379 Chr.17: 70133049-70133049 on Build GRCh38
rs545057636 Chr.17: 70133108-70133108 on Build GRCh38
rs1020054748 Chr.17: 70133051-70133051 on Build GRCh38
rs764497852 Chr.17: 70132950-70132950 on Build GRCh38
rs374457695 Chr.17: 70133144-70133144 on Build GRCh38
rs142625269 Chr.17: 70132991-70132991 on Build GRCh38
rs762095838 Chr.17: 70133088-70133088 on Build GRCh38
rs923043050 Chr.17: 70133116-70133116 on Build GRCh38

View Full Product Details