Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 120213973-120214216 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PXN
Entrez Gene ID
5829
Gene Name
paxillin
Gene Aliases
-
UniGene
Hs.446336
Gene Chromosome Location
Chr.12: 120210439-120265771 on Build GRCh38
Transcripts
NM_001080855.2 NM_001243756.1 NM_002859.3 NM_025157.4 XM_005253917.3 XM_006719532.1 XM_006719534.1 XM_006719535.3 XM_006719536.2 XM_011538622.1 XM_011538623.1 XM_017019726.1 XM_017019727.1 XM_017019728.1 XM_017019729.1 XM_017019730.1 XM_017019731.1 XM_017019733.1 XM_017019734.1 XM_017019735.1 XM_017019736.1 XM_017019737.1 XM_017019738.1 XM_017019739.1 XM_017019740.1 XM_017019741.1 XM_017019742.1 XM_017019743.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7417712 c.1199G>T p.R400L Chr.12: 120214195-120214195 on Build GRCh38
7417711 c.1301G>T p.R434L Chr.12: 120214195-120214195 on Build GRCh38
1152475 c.1356C>T p.P452P Chr.12: 120214140-120214140 on Build GRCh38
6298044 c.728C>A p.A243D Chr.12: 120214204-120214204 on Build GRCh38
6298042 c.1292C>A p.A431D Chr.12: 120214204-120214204 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756374374 Chr.12: 120214195-120214195 on Build GRCh38
rs759093168 Chr.12: 120214002-120214002 on Build GRCh38
rs980680895 Chr.12: 120214179-120214179 on Build GRCh38
rs960402847 Chr.12: 120214067-120214067 on Build GRCh38
rs540608652 Chr.12: 120214162-120214162 on Build GRCh38
rs898910607 Chr.12: 120214108-120214108 on Build GRCh38
rs778995962 Chr.12: 120214118-120214118 on Build GRCh38
rs749155386 Chr.12: 120214091-120214091 on Build GRCh38
rs372813036 Chr.12: 120214089-120214089 on Build GRCh38
rs200974822 Chr.12: 120214154-120214154 on Build GRCh38
rs752019807 Chr.12: 120214031-120214031 on Build GRCh38
rs746023566 Chr.12: 120214131-120214131 on Build GRCh38
rs923183684 Chr.12: 120214196-120214196 on Build GRCh38
rs750922909 Chr.12: 120214024-120214024 on Build GRCh38
rs980244760 Chr.12: 120214021-120214021 on Build GRCh38
rs781333003 Chr.12: 120214033-120214033 on Build GRCh38
rs749278232 Chr.12: 120213998-120213998 on Build GRCh38
rs770597404 Chr.12: 120213999-120213999 on Build GRCh38
rs766632483 Chr.12: 120214027-120214027 on Build GRCh38
rs369457359 Chr.12: 120214132-120214132 on Build GRCh38
rs1051578234 Chr.12: 120214081-120214081 on Build GRCh38
rs375047800 Chr.12: 120214110-120214110 on Build GRCh38
rs773082429 Chr.12: 120213992-120213992 on Build GRCh38
rs369071213 Chr.12: 120214026-120214026 on Build GRCh38
rs767264354 Chr.12: 120214003-120214003 on Build GRCh38
rs913790093 Chr.12: 120214076-120214077 on Build GRCh38
rs763722281 Chr.12: 120214014-120214014 on Build GRCh38
rs558454285 Chr.12: 120214140-120214140 on Build GRCh38
rs1033351105 Chr.12: 120214058-120214058 on Build GRCh38
rs775289495 Chr.12: 120214004-120214004 on Build GRCh38
rs777866965 Chr.12: 120214090-120214090 on Build GRCh38
rs964600835 Chr.12: 120214139-120214139 on Build GRCh38
rs1001754739 Chr.12: 120214043-120214043 on Build GRCh38
rs757277641 Chr.12: 120214000-120214000 on Build GRCh38
rs748389280 Chr.12: 120214039-120214039 on Build GRCh38
rs772010586 Chr.12: 120214113-120214113 on Build GRCh38
rs768251599 Chr.12: 120214121-120214121 on Build GRCh38
rs776330217 Chr.12: 120214122-120214122 on Build GRCh38
rs369639655 Chr.12: 120214079-120214079 on Build GRCh38
rs770955349 Chr.12: 120214095-120214095 on Build GRCh38
rs745613009 Chr.12: 120214106-120214106 on Build GRCh38
rs1013242382 Chr.12: 120214069-120214069 on Build GRCh38
rs375124475 Chr.12: 120214032-120214032 on Build GRCh38
rs775237469 Chr.12: 120214115-120214115 on Build GRCh38
rs1026085389 Chr.12: 120214088-120214088 on Build GRCh38
rs774009525 Chr.12: 120214105-120214105 on Build GRCh38
rs906129403 Chr.12: 120214038-120214038 on Build GRCh38
rs759893884 Chr.12: 120214153-120214153 on Build GRCh38

View Full Product Details