Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 50666106-50666379 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SHANK1
Entrez Gene ID
50944
Gene Name
SH3 and multiple ankyrin repeat domains 1
Gene Aliases
SPANK-1, SSTRIP, synamon
UniGene
Hs.274255
Gene Chromosome Location
Chr.19: 50659257-50719779 on Build GRCh38
Transcripts
NM_016148.3 XM_006723233.3 XM_011527013.2 XM_011527014.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4080454 c.3777T>C p.S1259S Chr.19: 50666344-50666344 on Build GRCh38
5004066 c.3807G>A p.G1269G Chr.19: 50666314-50666314 on Build GRCh38
7147026 c.5582C>T p.S1861L Chr.19: 50666378-50666378 on Build GRCh38
1712642 c.3866G>A p.G1289E Chr.19: 50666255-50666255 on Build GRCh38
6038248 c.3752C>A p.A1251D Chr.19: 50666369-50666369 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs199998427 Chr.19: 50666221-50666221 on Build GRCh38
rs566966888 Chr.19: 50666249-50666249 on Build GRCh38
rs200413731 Chr.19: 50666228-50666228 on Build GRCh38
rs760583150 Chr.19: 50666161-50666161 on Build GRCh38
rs1053704446 Chr.19: 50666125-50666125 on Build GRCh38
rs761631160 Chr.19: 50666318-50666318 on Build GRCh38
rs774314308 Chr.19: 50666239-50666239 on Build GRCh38
rs764656035 Chr.19: 50666211-50666211 on Build GRCh38
rs771405315 Chr.19: 50666237-50666237 on Build GRCh38
rs778008546 Chr.19: 50666288-50666288 on Build GRCh38
rs776419436 Chr.19: 50666257-50666257 on Build GRCh38
rs776315087 Chr.19: 50666317-50666317 on Build GRCh38
rs746633558 Chr.19: 50666226-50666226 on Build GRCh38
rs757212977 Chr.19: 50666210-50666210 on Build GRCh38
rs1011658644 Chr.19: 50666265-50666265 on Build GRCh38
rs772243664 Chr.19: 50666241-50666241 on Build GRCh38
rs772335528 Chr.19: 50666311-50666311 on Build GRCh38
rs754623779 Chr.19: 50666280-50666280 on Build GRCh38
rs780276977 Chr.19: 50666312-50666312 on Build GRCh38
rs942428865 Chr.19: 50666138-50666138 on Build GRCh38
rs549430068 Chr.19: 50666256-50666256 on Build GRCh38
rs370229569 Chr.19: 50666242-50666242 on Build GRCh38
rs201606756 Chr.19: 50666293-50666293 on Build GRCh38
rs1047443510 Chr.19: 50666199-50666199 on Build GRCh38
rs779510181 Chr.19: 50666305-50666305 on Build GRCh38
rs919787764 Chr.19: 50666162-50666162 on Build GRCh38
rs757732528 Chr.19: 50666296-50666296 on Build GRCh38
rs567195198 Chr.19: 50666259-50666259 on Build GRCh38
rs894526598 Chr.19: 50666248-50666248 on Build GRCh38
rs749935034 Chr.19: 50666271-50666271 on Build GRCh38
rs369466131 Chr.19: 50666143-50666143 on Build GRCh38
rs768863534 Chr.19: 50666314-50666314 on Build GRCh38
rs774711022 Chr.19: 50666290-50666298 on Build GRCh38
rs763958004 Chr.19: 50666185-50666185 on Build GRCh38
rs761668537 Chr.19: 50666258-50666258 on Build GRCh38
rs1046503378 Chr.19: 50666173-50666173 on Build GRCh38
rs778191544 Chr.19: 50666230-50666230 on Build GRCh38
rs374550090 Chr.19: 50666186-50666186 on Build GRCh38
rs192533917 Chr.19: 50666231-50666231 on Build GRCh38
rs751275163 Chr.19: 50666276-50666276 on Build GRCh38
rs745901446 Chr.19: 50666240-50666240 on Build GRCh38
rs549051295 Chr.19: 50666220-50666220 on Build GRCh38
rs779642061 Chr.19: 50666222-50666222 on Build GRCh38
rs945432764 Chr.19: 50666168-50666168 on Build GRCh38

View Full Product Details