Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 104002092-104002357 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLK
Entrez Gene ID
9748
Gene Name
STE20 like kinase
Gene Aliases
LOSK, STK2, bA16H23.1, se20-9
UniGene
Hs.591922
Gene Chromosome Location
Chr.10: 103967173-104029233 on Build GRCh38
Transcripts
NM_001304743.1 NM_014720.3 XM_011540401.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5836188 c.1037C>T p.A346V Chr.10: 104002215-104002215 on Build GRCh38
1266182 c.1133G>A p.S378N Chr.10: 104002311-104002311 on Build GRCh38
6577107 c.1156A>G p.T386A Chr.10: 104002334-104002334 on Build GRCh38
6268272 c.1036G>C p.A346P Chr.10: 104002214-104002214 on Build GRCh38
1345620 c.1077T>A p.I359I Chr.10: 104002255-104002255 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761701370 Chr.10: 104002190-104002190 on Build GRCh38
rs779798295 Chr.10: 104002140-104002140 on Build GRCh38
rs199730741 Chr.10: 104002295-104002295 on Build GRCh38
rs147685845 Chr.10: 104002177-104002177 on Build GRCh38
rs752774132 Chr.10: 104002274-104002274 on Build GRCh38
rs767850991 Chr.10: 104002298-104002299 on Build GRCh38
rs766599427 Chr.10: 104002205-104002205 on Build GRCh38
rs754064497 Chr.10: 104002213-104002213 on Build GRCh38
rs751988215 Chr.10: 104002284-104002284 on Build GRCh38
rs140715399 Chr.10: 104002283-104002283 on Build GRCh38
rs769578066 Chr.10: 104002136-104002136 on Build GRCh38
rs778306581 Chr.10: 104002132-104002132 on Build GRCh38
rs759799710 Chr.10: 104002214-104002214 on Build GRCh38
rs750250468 Chr.10: 104002301-104002306 on Build GRCh38
rs762157957 Chr.10: 104002227-104002228 on Build GRCh38
rs773019094 Chr.10: 104002195-104002195 on Build GRCh38
rs1048366320 Chr.10: 104002271-104002271 on Build GRCh38
rs544469688 Chr.10: 104002175-104002175 on Build GRCh38
rs143489434 Chr.10: 104002256-104002256 on Build GRCh38
rs754633596 Chr.10: 104002335-104002335 on Build GRCh38
rs760485209 Chr.10: 104002335-104002335 on Build GRCh38
rs781689932 Chr.10: 104002291-104002291 on Build GRCh38
rs757756541 Chr.10: 104002288-104002288 on Build GRCh38
rs965033276 Chr.10: 104002156-104002156 on Build GRCh38
rs765281537 Chr.10: 104002250-104002250 on Build GRCh38
rs768121994 Chr.10: 104002185-104002185 on Build GRCh38
rs773877151 Chr.10: 104002189-104002189 on Build GRCh38
rs748929327 Chr.10: 104002318-104002318 on Build GRCh38
rs201028021 Chr.10: 104002277-104002277 on Build GRCh38
rs962503769 Chr.10: 104002158-104002158 on Build GRCh38
rs373397561 Chr.10: 104002197-104002197 on Build GRCh38
rs979991855 Chr.10: 104002131-104002131 on Build GRCh38
rs201543977 Chr.10: 104002191-104002191 on Build GRCh38
rs373285860 Chr.10: 104002282-104002282 on Build GRCh38
rs141340729 Chr.10: 104002135-104002135 on Build GRCh38

View Full Product Details