Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 65027413-65027686 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARL2-SNX15
Entrez Gene ID
100528018
Gene Name
ARL2-SNX15 readthrough (NMD candidate)
Gene Aliases
-
UniGene
Hs.80132
Gene Chromosome Location
Chr.11: 65014113-65040572 on Build GRCh38
Transcripts
NR_037650.1 NM_013306.4 NM_147777.3

Gene Symbol
SNX15
Entrez Gene ID
29907
Gene Name
sorting nexin 15
Gene Aliases
HSAF001435
UniGene
Hs.80132
Gene Chromosome Location
Chr.11: 65027408-65040572 on Build GRCh38
Transcripts
NR_037650.1 NM_013306.4 NM_147777.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2164908 c.79G>A p.E27K Chr.11: 65027616-65027616 on Build GRCh38
6786415 c.49G>A p.D17N Chr.11: 65027586-65027586 on Build GRCh38
1227041 c.8G>A p.R3H Chr.11: 65027545-65027545 on Build GRCh38
5739307 c.69G>T p.K23N Chr.11: 65027606-65027606 on Build GRCh38
6069901 c.2T>C p.M1T Chr.11: 65027539-65027539 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200754789 Chr.11: 65027656-65027656 on Build GRCh38
rs370919667 Chr.11: 65027516-65027516 on Build GRCh38
rs374727432 Chr.11: 65027647-65027647 on Build GRCh38
rs776039886 Chr.11: 65027490-65027490 on Build GRCh38
rs780883862 Chr.11: 65027525-65027525 on Build GRCh38
rs369973827 Chr.11: 65027512-65027512 on Build GRCh38
rs534624602 Chr.11: 65027507-65027507 on Build GRCh38
rs545793141 Chr.11: 65027454-65027454 on Build GRCh38
rs772410080 Chr.11: 65027558-65027558 on Build GRCh38
rs775459814 Chr.11: 65027587-65027587 on Build GRCh38
rs201669379 Chr.11: 65027554-65027554 on Build GRCh38
rs749734356 Chr.11: 65027648-65027648 on Build GRCh38
rs369282650 Chr.11: 65027511-65027511 on Build GRCh38
rs763604711 Chr.11: 65027601-65027601 on Build GRCh38
rs768738863 Chr.11: 65027544-65027544 on Build GRCh38
rs773635231 Chr.11: 65027562-65027562 on Build GRCh38
rs765147382 Chr.11: 65027570-65027570 on Build GRCh38
rs568235874 Chr.11: 65027508-65027508 on Build GRCh38
rs755452996 Chr.11: 65027649-65027649 on Build GRCh38
rs148167643 Chr.11: 65027545-65027545 on Build GRCh38
rs751024202 Chr.11: 65027611-65027611 on Build GRCh38
rs767775453 Chr.11: 65027511-65027515 on Build GRCh38
rs767120653 Chr.11: 65027641-65027641 on Build GRCh38
rs376340866 Chr.11: 65027524-65027524 on Build GRCh38
rs768663767 Chr.11: 65027505-65027505 on Build GRCh38
rs761997851 Chr.11: 65027510-65027511 on Build GRCh38
rs376099592 Chr.11: 65027665-65027665 on Build GRCh38
rs892465414 Chr.11: 65027441-65027441 on Build GRCh38
rs375890503 Chr.11: 65027486-65027486 on Build GRCh38
rs759383006 Chr.11: 65027564-65027564 on Build GRCh38
rs756831824 Chr.11: 65027613-65027613 on Build GRCh38
rs3832791 Chr.11: 65027446-65027447 on Build GRCh38
rs750121081 Chr.11: 65027646-65027646 on Build GRCh38
rs770179913 Chr.11: 65027531-65027531 on Build GRCh38
rs1025028574 Chr.11: 65027476-65027476 on Build GRCh38
rs947511862 Chr.11: 65027438-65027438 on Build GRCh38
rs905356524 Chr.11: 65027480-65027480 on Build GRCh38
rs762805601 Chr.11: 65027597-65027597 on Build GRCh38
rs780513562 Chr.11: 65027535-65027535 on Build GRCh38
rs923486548 Chr.11: 65027666-65027666 on Build GRCh38
rs975799198 Chr.11: 65027549-65027549 on Build GRCh38
rs780117109 Chr.11: 65027550-65027550 on Build GRCh38
rs1043557331 Chr.11: 65027460-65027460 on Build GRCh38
rs1013600817 Chr.11: 65027452-65027452 on Build GRCh38
rs931801220 Chr.11: 65027654-65027654 on Build GRCh38
rs756344009 Chr.11: 65027481-65027481 on Build GRCh38
rs564214650 Chr.11: 65027542-65027542 on Build GRCh38
rs199754202 Chr.11: 65027519-65027519 on Build GRCh38
rs370769402 Chr.11: 65027526-65027526 on Build GRCh38
rs953980580 Chr.11: 65027484-65027484 on Build GRCh38
rs559279866 Chr.11: 65027488-65027488 on Build GRCh38
rs528772583 Chr.11: 65027492-65027492 on Build GRCh38

View Full Product Details