Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 31086762-31086989 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRSS53
Entrez Gene ID
339105
Gene Name
protease, serine 53
Gene Aliases
POL3S, UNQ308
UniGene
Hs.569575
Gene Chromosome Location
Chr.16: 31083424-31090035 on Build GRCh38
Transcripts
NM_001039503.2 XM_011545816.1 XM_011545817.1 XM_011545818.2 XM_011545819.1 XM_011545820.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3817860 c.287C>T p.S96F Chr.16: 31086854-31086854 on Build GRCh38
4718248 c.331delG p.V111fs*148 Chr.16: 31086810-31086810 on Build GRCh38
4718247 c.369C>T p.Y123Y Chr.16: 31086772-31086772 on Build GRCh38
3794815 c.319G>A p.E107K Chr.16: 31086822-31086822 on Build GRCh38
1377584 c.371G>A p.S124N Chr.16: 31086770-31086770 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs371524059 Chr.16: 31086823-31086823 on Build GRCh38
rs781325618 Chr.16: 31086889-31086889 on Build GRCh38
rs1051052208 Chr.16: 31086848-31086848 on Build GRCh38
rs761160521 Chr.16: 31086817-31086817 on Build GRCh38
rs768770514 Chr.16: 31086790-31086790 on Build GRCh38
rs772868279 Chr.16: 31086810-31086810 on Build GRCh38
rs1018466959 Chr.16: 31086917-31086917 on Build GRCh38
rs774182492 Chr.16: 31086936-31086936 on Build GRCh38
rs559983068 Chr.16: 31086845-31086845 on Build GRCh38
rs375934235 Chr.16: 31086793-31086793 on Build GRCh38
rs1043819134 Chr.16: 31086928-31086928 on Build GRCh38
rs780266645 Chr.16: 31086846-31086846 on Build GRCh38
rs138969193 Chr.16: 31086822-31086822 on Build GRCh38
rs754932389 Chr.16: 31086854-31086854 on Build GRCh38
rs769305312 Chr.16: 31086804-31086804 on Build GRCh38
rs200670079 Chr.16: 31086786-31086786 on Build GRCh38
rs765543793 Chr.16: 31086841-31086841 on Build GRCh38
rs770365663 Chr.16: 31086814-31086814 on Build GRCh38
rs777438043 Chr.16: 31086907-31086907 on Build GRCh38
rs748012747 Chr.16: 31086799-31086799 on Build GRCh38
rs534280793 Chr.16: 31086966-31086966 on Build GRCh38
rs746883770 Chr.16: 31086789-31086789 on Build GRCh38
rs748915939 Chr.16: 31086937-31086937 on Build GRCh38
rs770631427 Chr.16: 31086941-31086941 on Build GRCh38
rs377393908 Chr.16: 31086811-31086811 on Build GRCh38
rs762286967 Chr.16: 31086828-31086828 on Build GRCh38
rs746303464 Chr.16: 31086895-31086895 on Build GRCh38
rs770661511 Chr.16: 31086922-31086923 on Build GRCh38
rs768206883 Chr.16: 31086847-31086847 on Build GRCh38
rs748054376 Chr.16: 31086892-31086892 on Build GRCh38
rs74015064 Chr.16: 31086902-31086902 on Build GRCh38

View Full Product Details