Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 147653722-147653991 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRMT9
Entrez Gene ID
90826
Gene Name
protein arginine methyltransferase 9
Gene Aliases
PRMT10
UniGene
Hs.591692
Gene Chromosome Location
Chr.4: 147635626-147684230 on Build GRCh38
Transcripts
NM_001304458.1 NM_138364.3 XM_017008823.1 XM_017008824.1 XM_017008825.1 XM_017008826.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3600778 c.2041T>C p.S681P Chr.4: 147653856-147653856 on Build GRCh38
4717867 c.2004G>C p.G668G Chr.4: 147653893-147653893 on Build GRCh38
1618461 c.2033C>G p.A678G Chr.4: 147653864-147653864 on Build GRCh38
39730 c.1984G>A p.D662N Chr.4: 147653913-147653913 on Build GRCh38
6989488 c.2013G>A p.Q671Q Chr.4: 147653884-147653884 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1015127152 Chr.4: 147653822-147653822 on Build GRCh38
rs1053963742 Chr.4: 147653942-147653942 on Build GRCh38
rs538790926 Chr.4: 147653871-147653871 on Build GRCh38
rs138969454 Chr.4: 147653920-147653920 on Build GRCh38
rs767676759 Chr.4: 147653919-147653919 on Build GRCh38
rs932840710 Chr.4: 147653951-147653951 on Build GRCh38
rs779924735 Chr.4: 147653949-147653949 on Build GRCh38
rs370542261 Chr.4: 147653926-147653926 on Build GRCh38
rs923607161 Chr.4: 147653909-147653909 on Build GRCh38
rs200063176 Chr.4: 147653880-147653880 on Build GRCh38
rs80141801 Chr.4: 147653899-147653899 on Build GRCh38
rs184937144 Chr.4: 147653772-147653772 on Build GRCh38
rs773441385 Chr.4: 147653879-147653879 on Build GRCh38
rs936394458 Chr.4: 147653928-147653928 on Build GRCh38
rs772591492 Chr.4: 147653811-147653811 on Build GRCh38
rs760795444 Chr.4: 147653923-147653923 on Build GRCh38
rs890627316 Chr.4: 147653795-147653795 on Build GRCh38
rs199616123 Chr.4: 147653875-147653875 on Build GRCh38
rs768326884 Chr.4: 147653828-147653828 on Build GRCh38
rs368548509 Chr.4: 147653810-147653811 on Build GRCh38
rs139380764 Chr.4: 147653855-147653855 on Build GRCh38
rs758383196 Chr.4: 147653939-147653939 on Build GRCh38
rs142877020 Chr.4: 147653897-147653897 on Build GRCh38
rs748609949 Chr.4: 147653748-147653752 on Build GRCh38
rs747773557 Chr.4: 147653873-147653873 on Build GRCh38
rs990991506 Chr.4: 147653961-147653961 on Build GRCh38
rs758263957 Chr.4: 147653810-147653810 on Build GRCh38
rs914716907 Chr.4: 147653950-147653950 on Build GRCh38
rs779628896 Chr.4: 147653821-147653821 on Build GRCh38
rs977783806 Chr.4: 147653907-147653907 on Build GRCh38
rs556897380 Chr.4: 147653826-147653826 on Build GRCh38
rs750118595 Chr.4: 147653932-147653932 on Build GRCh38
rs763516738 Chr.4: 147653895-147653895 on Build GRCh38

View Full Product Details