Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 75794039-75794292 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RABGGTB
Entrez Gene ID
5876
Gene Name
Rab geranylgeranyltransferase beta subunit
Gene Aliases
GGTB
UniGene
Hs.78948
Gene Chromosome Location
Chr.1: 75786194-75795090 on Build GRCh38
Transcripts
NM_004582.3 NR_073562.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1222968 c.850G>T p.D284Y Chr.1: 75794228-75794228 on Build GRCh38
3948672 c.730T>G p.W244G Chr.1: 75794108-75794108 on Build GRCh38
682095 c.775G>T p.D259Y Chr.1: 75794153-75794153 on Build GRCh38
5342993 c.835G>A p.A279T Chr.1: 75794213-75794213 on Build GRCh38
912156 c.790C>T p.R264C Chr.1: 75794168-75794168 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374261688 Chr.1: 75794203-75794203 on Build GRCh38
rs1011574194 Chr.1: 75794128-75794128 on Build GRCh38
rs1802902 Chr.1: 75794159-75794159 on Build GRCh38
rs752724989 Chr.1: 75794198-75794198 on Build GRCh38
rs745412340 Chr.1: 75794232-75794232 on Build GRCh38
rs751104630 Chr.1: 75794207-75794207 on Build GRCh38
rs746718462 Chr.1: 75794252-75794253 on Build GRCh38
rs769343358 Chr.1: 75794238-75794238 on Build GRCh38
rs763206015 Chr.1: 75794125-75794125 on Build GRCh38
rs751775059 Chr.1: 75794155-75794157 on Build GRCh38
rs754317252 Chr.1: 75794156-75794159 on Build GRCh38
rs780632138 Chr.1: 75794231-75794231 on Build GRCh38
rs991203122 Chr.1: 75794256-75794256 on Build GRCh38
rs756946663 Chr.1: 75794226-75794226 on Build GRCh38
rs200508627 Chr.1: 75794252-75794252 on Build GRCh38
rs201388291 Chr.1: 75794162-75794162 on Build GRCh38
rs762102774 Chr.1: 75794146-75794146 on Build GRCh38
rs981262171 Chr.1: 75794202-75794202 on Build GRCh38
rs756448664 Chr.1: 75794168-75794168 on Build GRCh38
rs776017566 Chr.1: 75794122-75794122 on Build GRCh38
rs765231299 Chr.1: 75794180-75794180 on Build GRCh38
rs770306662 Chr.1: 75794074-75794074 on Build GRCh38
rs556286995 Chr.1: 75794095-75794095 on Build GRCh38
rs553836946 Chr.1: 75794185-75794185 on Build GRCh38
rs143074736 Chr.1: 75794169-75794169 on Build GRCh38
rs79193513 Chr.1: 75794242-75794242 on Build GRCh38
rs757854818 Chr.1: 75794203-75794205 on Build GRCh38
rs779614416 Chr.1: 75794204-75794205 on Build GRCh38
rs777687756 Chr.1: 75794206-75794206 on Build GRCh38
rs142398042 Chr.1: 75794151-75794151 on Build GRCh38
rs184984454 Chr.1: 75794140-75794140 on Build GRCh38
rs754657495 Chr.1: 75794264-75794266 on Build GRCh38
rs147978163 Chr.1: 75794132-75794132 on Build GRCh38
rs746316488 Chr.1: 75794070-75794070 on Build GRCh38
rs768837452 Chr.1: 75794255-75794255 on Build GRCh38

View Full Product Details