Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 12452288-12452555 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SPIRE1
Entrez Gene ID
56907
Gene Name
spire type actin nucleation factor 1
Gene Aliases
Spir-1
UniGene
Hs.515283
Gene Chromosome Location
Chr.18: 12446512-12658102 on Build GRCh38
Transcripts
NM_001128626.1 NM_001128627.1 NM_020148.2 XM_005258122.4 XM_011525701.1 XM_011525702.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5462747 c.1427G>T p.G476V Chr.18: 12452321-12452321 on Build GRCh38
5735084 c.1852G>T p.V618L Chr.18: 12452508-12452508 on Build GRCh38
5055949 c.1879C>T p.R627W Chr.18: 12452388-12452388 on Build GRCh38
7297495 c.1379A>G p.Y460C Chr.18: 12452369-12452369 on Build GRCh38
5853925 c.1460C>T p.T487I Chr.18: 12452288-12452288 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs568852514 Chr.18: 12452388-12452388 on Build GRCh38
rs755135923 Chr.18: 12452315-12452315 on Build GRCh38
rs745332264 Chr.18: 12452356-12452356 on Build GRCh38
rs765397535 Chr.18: 12452424-12452424 on Build GRCh38
rs865907652 Chr.18: 12452311-12452311 on Build GRCh38
rs533034976 Chr.18: 12452397-12452397 on Build GRCh38
rs748768107 Chr.18: 12452509-12452509 on Build GRCh38
rs137893351 Chr.18: 12452502-12452502 on Build GRCh38
rs566343540 Chr.18: 12452520-12452520 on Build GRCh38
rs747873118 Chr.18: 12452527-12452527 on Build GRCh38
rs145284625 Chr.18: 12452497-12452497 on Build GRCh38
rs201396204 Chr.18: 12452456-12452456 on Build GRCh38
rs367962520 Chr.18: 12452335-12452335 on Build GRCh38
rs746746069 Chr.18: 12452318-12452318 on Build GRCh38
rs767972169 Chr.18: 12452382-12452382 on Build GRCh38
rs770215686 Chr.18: 12452428-12452428 on Build GRCh38
rs142226113 Chr.18: 12452320-12452320 on Build GRCh38
rs748817192 Chr.18: 12452363-12452363 on Build GRCh38
rs775194626 Chr.18: 12452358-12452358 on Build GRCh38
rs774082973 Chr.18: 12452373-12452373 on Build GRCh38
rs762032609 Chr.18: 12452380-12452380 on Build GRCh38
rs768302986 Chr.18: 12452515-12452515 on Build GRCh38
rs781105971 Chr.18: 12452440-12452440 on Build GRCh38
rs370928358 Chr.18: 12452371-12452371 on Build GRCh38
rs772312287 Chr.18: 12452530-12452530 on Build GRCh38
rs769420628 Chr.18: 12452355-12452355 on Build GRCh38
rs375167237 Chr.18: 12452427-12452427 on Build GRCh38
rs200409524 Chr.18: 12452445-12452445 on Build GRCh38
rs779112520 Chr.18: 12452317-12452317 on Build GRCh38
rs754002404 Chr.18: 12452424-12452424 on Build GRCh38
rs773590464 Chr.18: 12452387-12452387 on Build GRCh38
rs756974253 Chr.18: 12452438-12452438 on Build GRCh38
rs745856037 Chr.18: 12452346-12452346 on Build GRCh38
rs753847935 Chr.18: 12452394-12452394 on Build GRCh38

View Full Product Details