Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 19772357-19772622 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PSPC1
Entrez Gene ID
55269
Gene Name
paraspeckle component 1
Gene Aliases
PSP1
UniGene
Hs.213198
Gene Chromosome Location
Chr.13: 19674752-19783047 on Build GRCh38
Transcripts
NM_001042414.2 NR_003272.2 NR_044998.1 XM_006719844.3 XM_011535137.1 XM_011535138.2 XM_011535139.1 XM_011535140.1 XM_011535141.2 XM_011535142.2 XM_017020650.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4046329 c.479T>C p.V160A Chr.13: 19772437-19772437 on Build GRCh38
2265096 c.454G>A p.A152T Chr.13: 19772462-19772462 on Build GRCh38
4046328 c.487C>T p.L163F Chr.13: 19772429-19772429 on Build GRCh38
5457814 c.373-1G>A p.? Chr.13: 19772544-19772544 on Build GRCh38
945788 c.504C>T p.S168S Chr.13: 19772412-19772412 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762169666 Chr.13: 19772586-19772587 on Build GRCh38
rs77006383 Chr.13: 19772502-19772502 on Build GRCh38
rs574996931 Chr.13: 19772552-19772552 on Build GRCh38
rs763830985 Chr.13: 19772475-19772475 on Build GRCh38
rs778971748 Chr.13: 19772542-19772542 on Build GRCh38
rs770697710 Chr.13: 19772445-19772445 on Build GRCh38
rs746779056 Chr.13: 19772439-19772439 on Build GRCh38
rs542120487 Chr.13: 19772600-19772600 on Build GRCh38
rs780203109 Chr.13: 19772580-19772580 on Build GRCh38
rs983979731 Chr.13: 19772477-19772477 on Build GRCh38
rs537047009 Chr.13: 19772436-19772436 on Build GRCh38
rs201903737 Chr.13: 19772497-19772497 on Build GRCh38
rs745772378 Chr.13: 19772463-19772463 on Build GRCh38
rs752911035 Chr.13: 19772549-19772549 on Build GRCh38
rs775115126 Chr.13: 19772473-19772473 on Build GRCh38
rs375378380 Chr.13: 19772434-19772434 on Build GRCh38
rs541595269 Chr.13: 19772505-19772505 on Build GRCh38
rs377193819 Chr.13: 19772551-19772551 on Build GRCh38
rs745696275 Chr.13: 19772557-19772557 on Build GRCh38
rs769889904 Chr.13: 19772470-19772470 on Build GRCh38
rs762606545 Chr.13: 19772474-19772474 on Build GRCh38
rs907860362 Chr.13: 19772481-19772481 on Build GRCh38
rs61159938 Chr.13: 19772433-19772433 on Build GRCh38
rs373146127 Chr.13: 19772532-19772532 on Build GRCh38
rs867206225 Chr.13: 19772492-19772492 on Build GRCh38
rs559723605 Chr.13: 19772529-19772529 on Build GRCh38
rs756583285 Chr.13: 19772511-19772511 on Build GRCh38
rs766922316 Chr.13: 19772513-19772513 on Build GRCh38
rs776306907 Chr.13: 19772454-19772454 on Build GRCh38
rs200482553 Chr.13: 19772403-19772403 on Build GRCh38
rs763615108 Chr.13: 19772420-19772420 on Build GRCh38
rs755170644 Chr.13: 19772424-19772424 on Build GRCh38
rs201955578 Chr.13: 19772531-19772531 on Build GRCh38
rs749241049 Chr.13: 19772594-19772594 on Build GRCh38
rs769836902 Chr.13: 19772564-19772564 on Build GRCh38
rs773985876 Chr.13: 19772480-19772480 on Build GRCh38

View Full Product Details