Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 131947619-131947852 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PPP2R2D
Entrez Gene ID
55844
Gene Name
protein phosphatase 2 regulatory subunit Bdelta
Gene Aliases
MDS026
UniGene
Hs.380372
Gene Chromosome Location
Chr.10: 131900644-131971535 on Build GRCh38
Transcripts
XM_006717914.3 XM_017016407.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6455081 c.938A>C p.E313A Chr.10: 131947740-131947740 on Build GRCh38
6997999 c.888G>C p.E296D Chr.10: 131947690-131947690 on Build GRCh38
284195 c.914G>A p.R305H Chr.10: 131947716-131947716 on Build GRCh38
916294 c.983C>T p.S328L Chr.10: 131947785-131947785 on Build GRCh38
273144 c.817C>A p.R273R Chr.10: 131947619-131947619 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781904546 Chr.10: 131947705-131947705 on Build GRCh38
rs782767004 Chr.10: 131947695-131947695 on Build GRCh38
rs34473884 Chr.10: 131947781-131947781 on Build GRCh38
rs369435052 Chr.10: 131947706-131947706 on Build GRCh38
rs375481282 Chr.10: 131947696-131947696 on Build GRCh38
rs781897599 Chr.10: 131947809-131947809 on Build GRCh38
rs142846802 Chr.10: 131947785-131947785 on Build GRCh38
rs782046497 Chr.10: 131947664-131947664 on Build GRCh38
rs76274320 Chr.10: 131947649-131947649 on Build GRCh38
rs781924750 Chr.10: 131947780-131947780 on Build GRCh38
rs193198132 Chr.10: 131947716-131947716 on Build GRCh38
rs782395807 Chr.10: 131947647-131947647 on Build GRCh38
rs779401090 Chr.10: 131947825-131947825 on Build GRCh38
rs547732352 Chr.10: 131947715-131947715 on Build GRCh38
rs782085237 Chr.10: 131947765-131947765 on Build GRCh38
rs183379034 Chr.10: 131947819-131947819 on Build GRCh38
rs782471400 Chr.10: 131947698-131947698 on Build GRCh38
rs781799098 Chr.10: 131947803-131947803 on Build GRCh38
rs372077312 Chr.10: 131947684-131947684 on Build GRCh38
rs782804176 Chr.10: 131947681-131947681 on Build GRCh38
rs782742409 Chr.10: 131947802-131947802 on Build GRCh38
rs367744151 Chr.10: 131947651-131947651 on Build GRCh38
rs782691603 Chr.10: 131947699-131947699 on Build GRCh38
rs781875794 Chr.10: 131947797-131947797 on Build GRCh38
rs201790987 Chr.10: 131947726-131947726 on Build GRCh38
rs910373303 Chr.10: 131947808-131947808 on Build GRCh38
rs369446279 Chr.10: 131947786-131947786 on Build GRCh38
rs782559592 Chr.10: 131947805-131947805 on Build GRCh38
rs1012656436 Chr.10: 131947645-131947645 on Build GRCh38
rs782274738 Chr.10: 131947735-131947735 on Build GRCh38
rs782120311 Chr.10: 131947798-131947798 on Build GRCh38
rs549446500 Chr.10: 131947818-131947818 on Build GRCh38
rs782521857 Chr.10: 131947813-131947813 on Build GRCh38
rs781971947 Chr.10: 131947737-131947737 on Build GRCh38
rs782651908 Chr.10: 131947708-131947708 on Build GRCh38
rs782027193 Chr.10: 131947728-131947728 on Build GRCh38
rs782184502 Chr.10: 131947831-131947831 on Build GRCh38
rs782435865 Chr.10: 131947833-131947833 on Build GRCh38
rs1039762319 Chr.10: 131947822-131947822 on Build GRCh38
rs782713608 Chr.10: 131947768-131947768 on Build GRCh38
rs781981250 Chr.10: 131947648-131947648 on Build GRCh38
rs782121870 Chr.10: 131947689-131947689 on Build GRCh38
rs782190671 Chr.10: 131947720-131947720 on Build GRCh38

View Full Product Details