Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 47915182-47915421 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF81
Entrez Gene ID
347344
Gene Name
zinc finger protein 81
Gene Aliases
HFZ20, MRX45, dJ54B20.6
UniGene
Hs.114246
Gene Chromosome Location
Chr.X: 47836864-47925626 on Build GRCh38
Transcripts
NM_007137.3 XM_005272600.3 XM_011543899.2 XM_011543900.2 XM_017029486.1 XM_017029487.1

SNP Detail

SNP ID Chromosome Location
rs782560420 Chr.X: 47915354-47915354 on Build GRCh38
rs782029613 Chr.X: 47915299-47915299 on Build GRCh38
rs782742839 Chr.X: 47915353-47915353 on Build GRCh38
rs782282759 Chr.X: 47915388-47915388 on Build GRCh38
rs140856764 Chr.X: 47915308-47915308 on Build GRCh38
rs782274841 Chr.X: 47915237-47915237 on Build GRCh38
rs782687692 Chr.X: 47915370-47915370 on Build GRCh38
rs782692938 Chr.X: 47915295-47915295 on Build GRCh38
rs979194990 Chr.X: 47915210-47915210 on Build GRCh38
rs781785337 Chr.X: 47915338-47915338 on Build GRCh38
rs887268121 Chr.X: 47915362-47915362 on Build GRCh38
rs189912680 Chr.X: 47915355-47915355 on Build GRCh38
rs267606463 Chr.X: 47915268-47915268 on Build GRCh38
rs201362916 Chr.X: 47915213-47915213 on Build GRCh38
rs969211913 Chr.X: 47915209-47915209 on Build GRCh38
rs782182289 Chr.X: 47915242-47915242 on Build GRCh38
rs537825 Chr.X: 47915284-47915284 on Build GRCh38
rs782460656 Chr.X: 47915302-47915302 on Build GRCh38
rs371947038 Chr.X: 47915263-47915263 on Build GRCh38
rs199990232 Chr.X: 47915248-47915248 on Build GRCh38
rs782478712 Chr.X: 47915345-47915345 on Build GRCh38
rs781979833 Chr.X: 47915287-47915287 on Build GRCh38
rs373300904 Chr.X: 47915273-47915273 on Build GRCh38
rs1039488608 Chr.X: 47915347-47915347 on Build GRCh38
rs375487524 Chr.X: 47915220-47915220 on Build GRCh38
rs782090646 Chr.X: 47915288-47915288 on Build GRCh38
rs782263364 Chr.X: 47915271-47915271 on Build GRCh38

View Full Product Details