Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 44343025-44343298 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SPATS1
Entrez Gene ID
221409
Gene Name
spermatogenesis associated serine rich 1
Gene Aliases
DDIP, SPATA8, SRSP1
UniGene
Hs.135283
Gene Chromosome Location
Chr.6: 44342660-44377167 on Build GRCh38
Transcripts
NM_145026.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6471247 c.67_69delACC p.T23delT Chr.6: 44343162-44343164 on Build GRCh38
3627837 c.46C>T p.L16F Chr.6: 44343141-44343141 on Build GRCh38
3627839 c.53C>T p.S18F Chr.6: 44343148-44343148 on Build GRCh38
6884511 c.91G>C p.V31L Chr.6: 44343186-44343186 on Build GRCh38
367422 c.121G>A p.E41K Chr.6: 44343216-44343216 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs546746044 Chr.6: 44343144-44343144 on Build GRCh38
rs755444993 Chr.6: 44343221-44343221 on Build GRCh38
rs750805977 Chr.6: 44343119-44343119 on Build GRCh38
rs760546925 Chr.6: 44343163-44343163 on Build GRCh38
rs776569375 Chr.6: 44343189-44343189 on Build GRCh38
rs746892567 Chr.6: 44343256-44343256 on Build GRCh38
rs868723898 Chr.6: 44343110-44343110 on Build GRCh38
rs765536160 Chr.6: 44343194-44343194 on Build GRCh38
rs764105715 Chr.6: 44343106-44343106 on Build GRCh38
rs946940643 Chr.6: 44343118-44343118 on Build GRCh38
rs770701876 Chr.6: 44343081-44343081 on Build GRCh38
rs776680682 Chr.6: 44343265-44343265 on Build GRCh38
rs370149642 Chr.6: 44343274-44343274 on Build GRCh38
rs758794030 Chr.6: 44343209-44343209 on Build GRCh38
rs10948132 Chr.6: 44343117-44343117 on Build GRCh38
rs754037332 Chr.6: 44343111-44343111 on Build GRCh38
rs758884220 Chr.6: 44343133-44343133 on Build GRCh38
rs374573932 Chr.6: 44343271-44343271 on Build GRCh38
rs538272131 Chr.6: 44343148-44343148 on Build GRCh38
rs779832625 Chr.6: 44343248-44343248 on Build GRCh38
rs747548099 Chr.6: 44343137-44343137 on Build GRCh38
rs575454362 Chr.6: 44343214-44343214 on Build GRCh38
rs536003825 Chr.6: 44343222-44343222 on Build GRCh38
rs755599982 Chr.6: 44343139-44343139 on Build GRCh38
rs764172774 Chr.6: 44343186-44343186 on Build GRCh38
rs760769920 Chr.6: 44343104-44343104 on Build GRCh38
rs370511457 Chr.6: 44343088-44343088 on Build GRCh38
rs373023494 Chr.6: 44343092-44343092 on Build GRCh38
rs762011543 Chr.6: 44343191-44343191 on Build GRCh38
rs745544556 Chr.6: 44343234-44343234 on Build GRCh38
rs771800489 Chr.6: 44343245-44343245 on Build GRCh38
rs780677764 Chr.6: 44343134-44343134 on Build GRCh38
rs557194939 Chr.6: 44343200-44343200 on Build GRCh38
rs571680001 Chr.6: 44343228-44343228 on Build GRCh38
rs768465282 Chr.6: 44343264-44343264 on Build GRCh38
rs775361180 Chr.6: 44343161-44343161 on Build GRCh38
rs747871855 Chr.6: 44343247-44343247 on Build GRCh38
rs771888446 Chr.6: 44343158-44343158 on Build GRCh38
rs757429284 Chr.6: 44343115-44343115 on Build GRCh38
rs143387848 Chr.6: 44343146-44343146 on Build GRCh38
rs748731110 Chr.6: 44343226-44343226 on Build GRCh38
rs775453207 Chr.6: 44343101-44343101 on Build GRCh38
rs368991130 Chr.6: 44343087-44343087 on Build GRCh38
rs752046258 Chr.6: 44343215-44343215 on Build GRCh38
rs565777182 Chr.6: 44343154-44343154 on Build GRCh38
rs368940680 Chr.6: 44343273-44343273 on Build GRCh38
rs139954152 Chr.6: 44343143-44343143 on Build GRCh38
rs748914859 Chr.6: 44343063-44343063 on Build GRCh38

View Full Product Details