Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 34880032-34880303 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TAF11
Entrez Gene ID
6882
Gene Name
TATA-box binding protein associated factor 11
Gene Aliases
MGC:15243, PRO2134, TAF2I, TAFII28
UniGene
Hs.112444
Gene Chromosome Location
Chr.6: 34877778-34888071 on Build GRCh38
Transcripts
NM_001270488.1 NM_005643.3 XM_011514827.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7158169 c.408+2T>C p.? Chr.6: 34880287-34880287 on Build GRCh38
6273811 c.409-4C>A p.? Chr.6: 34880067-34880067 on Build GRCh38
7192912 c.416A>T p.Q139L Chr.6: 34880056-34880056 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs572580887 Chr.6: 34880173-34880173 on Build GRCh38
rs533757339 Chr.6: 34880115-34880115 on Build GRCh38
rs374201926 Chr.6: 34880263-34880263 on Build GRCh38
rs368708305 Chr.6: 34880278-34880278 on Build GRCh38
rs901878236 Chr.6: 34880219-34880219 on Build GRCh38
rs1006369482 Chr.6: 34880198-34880198 on Build GRCh38
rs749490510 Chr.6: 34880281-34880281 on Build GRCh38
rs879182986 Chr.6: 34880165-34880166 on Build GRCh38
rs997605999 Chr.6: 34880221-34880221 on Build GRCh38
rs748826119 Chr.6: 34880274-34880274 on Build GRCh38
rs774868760 Chr.6: 34880066-34880066 on Build GRCh38
rs187158837 Chr.6: 34880254-34880254 on Build GRCh38
rs751420558 Chr.6: 34880269-34880269 on Build GRCh38
rs756297082 Chr.6: 34880271-34880271 on Build GRCh38
rs896690778 Chr.6: 34880110-34880110 on Build GRCh38
rs993761727 Chr.6: 34880224-34880224 on Build GRCh38
rs546420779 Chr.6: 34880077-34880077 on Build GRCh38
rs777529509 Chr.6: 34880264-34880264 on Build GRCh38
rs560383660 Chr.6: 34880083-34880083 on Build GRCh38
rs764201662 Chr.6: 34880085-34880085 on Build GRCh38
rs764685922 Chr.6: 34880255-34880255 on Build GRCh38
rs1025268678 Chr.6: 34880250-34880250 on Build GRCh38
rs560981548 Chr.6: 34880237-34880237 on Build GRCh38
rs766176770 Chr.6: 34880096-34880096 on Build GRCh38
rs529217183 Chr.6: 34880244-34880244 on Build GRCh38
rs1016639310 Chr.6: 34880204-34880204 on Build GRCh38
rs367863055 Chr.6: 34880086-34880086 on Build GRCh38
rs769021125 Chr.6: 34880052-34880052 on Build GRCh38
rs762253297 Chr.6: 34880074-34880074 on Build GRCh38
rs4646939 Chr.6: 34880209-34880209 on Build GRCh38
rs1006833350 Chr.6: 34880098-34880098 on Build GRCh38
rs891548734 Chr.6: 34880063-34880063 on Build GRCh38
rs963906755 Chr.6: 34880276-34880276 on Build GRCh38

View Full Product Details