Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 21856597-21856836 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MBTPS2
Entrez Gene ID
51360
Gene Name
membrane bound transcription factor peptidase, site 2
Gene Aliases
BRESEK, IFAP, KFSD, KFSDX, OLMSX, S2P
UniGene
Hs.443490
Gene Chromosome Location
Chr.X: 21839538-21885423 on Build GRCh38
Transcripts
NM_015884.3 NM_206923.3

Gene Symbol
YY2
Entrez Gene ID
404281
Gene Name
YY2 transcription factor
Gene Aliases
ZNF631
UniGene
Hs.673601
Gene Chromosome Location
Chr.X: 21855987-21858727 on Build GRCh38
Transcripts
NM_015884.3 NM_206923.3

SNP Detail

SNP ID Chromosome Location
rs773669593 Chr.X: 21856676-21856676 on Build GRCh38
rs765855533 Chr.X: 21856753-21856753 on Build GRCh38
rs950117940 Chr.X: 21856658-21856658 on Build GRCh38
rs781656677 Chr.X: 21856808-21856808 on Build GRCh38
rs776472428 Chr.X: 21856684-21856684 on Build GRCh38
rs981417085 Chr.X: 21856661-21856661 on Build GRCh38
rs780575312 Chr.X: 21856659-21856659 on Build GRCh38
rs755435591 Chr.X: 21856800-21856800 on Build GRCh38
rs947717850 Chr.X: 21856727-21856727 on Build GRCh38
rs767388558 Chr.X: 21856767-21856790 on Build GRCh38
rs917826793 Chr.X: 21856654-21856654 on Build GRCh38
rs772729362 Chr.X: 21856700-21856700 on Build GRCh38
rs141654651 Chr.X: 21856691-21856691 on Build GRCh38
rs939212904 Chr.X: 21856782-21856782 on Build GRCh38
rs914761589 Chr.X: 21856736-21856736 on Build GRCh38
rs759382433 Chr.X: 21856754-21856754 on Build GRCh38
rs147081325 Chr.X: 21856791-21856791 on Build GRCh38
rs758407096 Chr.X: 21856793-21856793 on Build GRCh38
rs767275737 Chr.X: 21856768-21856768 on Build GRCh38
rs772576056 Chr.X: 21856655-21856655 on Build GRCh38
rs766237007 Chr.X: 21856722-21856722 on Build GRCh38
rs761498141 Chr.X: 21856685-21856685 on Build GRCh38
rs746388847 Chr.X: 21856635-21856635 on Build GRCh38
rs762811083 Chr.X: 21856706-21856706 on Build GRCh38
rs754095870 Chr.X: 21856769-21856769 on Build GRCh38
rs991177863 Chr.X: 21856623-21856623 on Build GRCh38
rs1054543788 Chr.X: 21856730-21856730 on Build GRCh38
rs375625645 Chr.X: 21856796-21856796 on Build GRCh38
rs765612557 Chr.X: 21856627-21856627 on Build GRCh38
rs914919324 Chr.X: 21856717-21856717 on Build GRCh38
rs751090496 Chr.X: 21856787-21856787 on Build GRCh38
rs749130176 Chr.X: 21856620-21856621 on Build GRCh38

View Full Product Details