Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 51699363-51699636 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TEX264
Entrez Gene ID
51368
Gene Name
testis expressed 264
Gene Aliases
ZSIG11
UniGene
Hs.517864
Gene Chromosome Location
Chr.3: 51671175-51704323 on Build GRCh38
Transcripts
NM_001129884.2 NM_001243725.1 NM_001243726.1 NM_001243727.2 NM_001278195.1 NM_015926.5 NR_024012.2 NR_103462.1 NR_103463.1 XM_006713195.3 XM_006713197.3 XM_006713198.2 XM_011533805.2 XM_017006573.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1046734 c.555A>G p.A185A Chr.3: 51699480-51699480 on Build GRCh38
7274141 c.633C>T p.T211T Chr.3: 51699558-51699558 on Build GRCh38
4733219 c.605G>T p.W202L Chr.3: 51699530-51699530 on Build GRCh38
7117675 c.582G>A p.E194E Chr.3: 51699507-51699507 on Build GRCh38
3427730 c.607C>T p.R203W Chr.3: 51699532-51699532 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs371777255 Chr.3: 51699456-51699456 on Build GRCh38
rs566855739 Chr.3: 51699605-51699605 on Build GRCh38
rs759970460 Chr.3: 51699558-51699558 on Build GRCh38
rs761097253 Chr.3: 51699410-51699410 on Build GRCh38
rs765560396 Chr.3: 51699432-51699432 on Build GRCh38
rs749215563 Chr.3: 51699486-51699486 on Build GRCh38
rs371011006 Chr.3: 51699482-51699482 on Build GRCh38
rs760505589 Chr.3: 51699587-51699587 on Build GRCh38
rs767253995 Chr.3: 51699581-51699581 on Build GRCh38
rs766844593 Chr.3: 51699415-51699415 on Build GRCh38
rs747356480 Chr.3: 51699552-51699552 on Build GRCh38
rs867158699 Chr.3: 51699549-51699549 on Build GRCh38
rs778012647 Chr.3: 51699534-51699534 on Build GRCh38
rs773727438 Chr.3: 51699572-51699572 on Build GRCh38
rs1051796530 Chr.3: 51699575-51699575 on Build GRCh38
rs778874112 Chr.3: 51699496-51699496 on Build GRCh38
rs773693745 Chr.3: 51699597-51699597 on Build GRCh38
rs751163484 Chr.3: 51699453-51699453 on Build GRCh38
rs141178453 Chr.3: 51699555-51699555 on Build GRCh38
rs909703663 Chr.3: 51699474-51699474 on Build GRCh38
rs201623991 Chr.3: 51699588-51699588 on Build GRCh38
rs752903364 Chr.3: 51699605-51699606 on Build GRCh38
rs1024889737 Chr.3: 51699516-51699516 on Build GRCh38
rs753054411 Chr.3: 51699533-51699533 on Build GRCh38
rs377591386 Chr.3: 51699564-51699564 on Build GRCh38
rs375657144 Chr.3: 51699610-51699610 on Build GRCh38
rs200150485 Chr.3: 51699430-51699430 on Build GRCh38
rs181163364 Chr.3: 51699383-51699383 on Build GRCh38
rs753751256 Chr.3: 51699592-51699592 on Build GRCh38
rs148617571 Chr.3: 51699481-51699481 on Build GRCh38
rs766230594 Chr.3: 51699530-51699532 on Build GRCh38
rs772366823 Chr.3: 51699396-51699396 on Build GRCh38
rs111793502 Chr.3: 51699577-51699577 on Build GRCh38
rs771311925 Chr.3: 51699554-51699554 on Build GRCh38
rs758162494 Chr.3: 51699386-51699386 on Build GRCh38
rs779992812 Chr.3: 51699485-51699485 on Build GRCh38
rs773556748 Chr.3: 51699409-51699409 on Build GRCh38
rs762481142 Chr.3: 51699431-51699431 on Build GRCh38
rs1006029295 Chr.3: 51699455-51699455 on Build GRCh38
rs756953605 Chr.3: 51699465-51699465 on Build GRCh38
rs948580613 Chr.3: 51699445-51699445 on Build GRCh38
rs969044528 Chr.3: 51699539-51699539 on Build GRCh38
rs755019035 Chr.3: 51699493-51699493 on Build GRCh38
rs748319268 Chr.3: 51699508-51699508 on Build GRCh38
rs748478449 Chr.3: 51699497-51699497 on Build GRCh38
rs756306461 Chr.3: 51699606-51699607 on Build GRCh38
rs1050481199 Chr.3: 51699405-51699405 on Build GRCh38
rs201087062 Chr.3: 51699394-51699394 on Build GRCh38

View Full Product Details