Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 179653275-179653522 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
USP13
Entrez Gene ID
8975
Gene Name
ubiquitin specific peptidase 13 (isopeptidase T-3)
Gene Aliases
ISOT3, IsoT-3
UniGene
Hs.175322
Gene Chromosome Location
Chr.3: 179653145-179789401 on Build GRCh38
Transcripts
NM_003940.2 XM_011513269.1 XM_017007426.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4157451 c.141C>T p.N47N Chr.3: 179653366-179653366 on Build GRCh38
5443058 c.164C>T p.S55F Chr.3: 179653389-179653389 on Build GRCh38
5642578 c.124G>T p.D42Y Chr.3: 179653349-179653349 on Build GRCh38
1231959 c.130G>A p.V44I Chr.3: 179653355-179653355 on Build GRCh38
1308850 c.72C>T p.I24I Chr.3: 179653297-179653297 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs570604627 Chr.3: 179653393-179653393 on Build GRCh38
rs950538623 Chr.3: 179653503-179653503 on Build GRCh38
rs1051215718 Chr.3: 179653458-179653458 on Build GRCh38
rs147844491 Chr.3: 179653320-179653320 on Build GRCh38
rs766266327 Chr.3: 179653378-179653378 on Build GRCh38
rs998394053 Chr.3: 179653501-179653501 on Build GRCh38
rs1038627410 Chr.3: 179653485-179653485 on Build GRCh38
rs372813389 Chr.3: 179653354-179653354 on Build GRCh38
rs1037326884 Chr.3: 179653478-179653478 on Build GRCh38
rs925927683 Chr.3: 179653391-179653391 on Build GRCh38
rs376341612 Chr.3: 179653389-179653389 on Build GRCh38
rs202069129 Chr.3: 179653394-179653394 on Build GRCh38
rs757684032 Chr.3: 179653442-179653442 on Build GRCh38
rs913317850 Chr.3: 179653452-179653452 on Build GRCh38
rs555102613 Chr.3: 179653464-179653464 on Build GRCh38
rs936040785 Chr.3: 179653420-179653420 on Build GRCh38
rs144481641 Chr.3: 179653300-179653300 on Build GRCh38
rs779051347 Chr.3: 179653295-179653295 on Build GRCh38
rs937653665 Chr.3: 179653463-179653463 on Build GRCh38
rs954172080 Chr.3: 179653384-179653384 on Build GRCh38
rs913728166 Chr.3: 179653437-179653437 on Build GRCh38
rs368463855 Chr.3: 179653407-179653407 on Build GRCh38
rs556049989 Chr.3: 179653444-179653444 on Build GRCh38
rs762749034 Chr.3: 179653375-179653375 on Build GRCh38
rs754170471 Chr.3: 179653433-179653433 on Build GRCh38
rs917147961 Chr.3: 179653322-179653322 on Build GRCh38
rs575887154 Chr.3: 179653445-179653445 on Build GRCh38
rs541447591 Chr.3: 179653453-179653453 on Build GRCh38
rs779215084 Chr.3: 179653443-179653443 on Build GRCh38
rs1005800601 Chr.3: 179653474-179653474 on Build GRCh38
rs778900244 Chr.3: 179653493-179653493 on Build GRCh38
rs2276803 Chr.3: 179653366-179653366 on Build GRCh38
rs764598147 Chr.3: 179653414-179653414 on Build GRCh38
rs888669336 Chr.3: 179653471-179653471 on Build GRCh38
rs1026024685 Chr.3: 179653502-179653502 on Build GRCh38
rs772176590 Chr.3: 179653316-179653316 on Build GRCh38
rs371148605 Chr.3: 179653466-179653466 on Build GRCh38
rs372026666 Chr.3: 179653413-179653413 on Build GRCh38
rs994918985 Chr.3: 179653497-179653497 on Build GRCh38
rs748058476 Chr.3: 179653332-179653332 on Build GRCh38
rs980045471 Chr.3: 179653385-179653385 on Build GRCh38
rs1007075983 Chr.3: 179653465-179653465 on Build GRCh38
rs781272923 Chr.3: 179653305-179653305 on Build GRCh38

View Full Product Details