Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 149611288-149611529 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM185A
Entrez Gene ID
84548
Gene Name
transmembrane protein 185A
Gene Aliases
CXorf13, FAM11A, FRAXF, ee3
UniGene
Hs.522172
Gene Chromosome Location
Chr.X: 149596556-149631912 on Build GRCh38
Transcripts
NM_001174092.2 NM_001282302.1 NM_032508.3 NR_104121.1

SNP Detail

SNP ID Chromosome Location
rs782352745 Chr.X: 149611344-149611344 on Build GRCh38
rs376945268 Chr.X: 149611464-149611464 on Build GRCh38
rs899583915 Chr.X: 149611345-149611345 on Build GRCh38
rs9887071 Chr.X: 149611481-149611481 on Build GRCh38
rs781888304 Chr.X: 149611395-149611395 on Build GRCh38
rs782405567 Chr.X: 149611348-149611348 on Build GRCh38
rs782191605 Chr.X: 149611347-149611347 on Build GRCh38
rs200479899 Chr.X: 149611354-149611354 on Build GRCh38
rs781974429 Chr.X: 149611359-149611359 on Build GRCh38
rs782438408 Chr.X: 149611399-149611399 on Build GRCh38
rs781875948 Chr.X: 149611458-149611458 on Build GRCh38
rs782554276 Chr.X: 149611346-149611346 on Build GRCh38
rs782730273 Chr.X: 149611377-149611377 on Build GRCh38
rs782175116 Chr.X: 149611411-149611411 on Build GRCh38
rs782114799 Chr.X: 149611351-149611351 on Build GRCh38
rs782695633 Chr.X: 149611336-149611336 on Build GRCh38
rs782768765 Chr.X: 149611439-149611439 on Build GRCh38
rs782050026 Chr.X: 149611363-149611363 on Build GRCh38
rs998520435 Chr.X: 149611470-149611470 on Build GRCh38
rs373599378 Chr.X: 149611349-149611349 on Build GRCh38

View Full Product Details