Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 99035067-99035328 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TSGA10
Entrez Gene ID
80705
Gene Name
testis specific 10
Gene Aliases
CEP4L, CT79
UniGene
Hs.120267
Gene Chromosome Location
Chr.2: 98997261-99154962 on Build GRCh38
Transcripts
NM_025244.2 NM_182911.3 XM_005264023.1 XM_005264024.2 XM_005264025.1 XM_005264026.1 XM_005264027.1 XM_005264029.1 XM_006712781.2 XM_011511934.1 XM_011511935.2 XM_011511937.1 XM_011511939.1 XM_011511940.2 XM_011511941.1 XM_011511942.1 XM_011511943.2 XM_017005035.1 XM_017005036.1 XM_017005037.1 XM_017005038.1 XM_017005039.1 XM_017005040.1 XM_017005041.1 XM_017005042.1 XM_017005043.1 XM_017005044.1 XM_017005045.1 XM_017005046.1 XM_017005047.1 XM_017005048.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1410245 c.1592C>T p.A531V Chr.2: 99035252-99035252 on Build GRCh38
1410244 c.1614+2T>G p.? Chr.2: 99035228-99035228 on Build GRCh38
5863068 c.1535G>A p.R512K Chr.2: 99035309-99035309 on Build GRCh38
1024300 c.1603G>T p.E535* Chr.2: 99035241-99035241 on Build GRCh38
1024299 c.1609G>T p.E537* Chr.2: 99035235-99035235 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs941027475 Chr.2: 99035203-99035203 on Build GRCh38
rs957435289 Chr.2: 99035141-99035141 on Build GRCh38
rs767942925 Chr.2: 99035231-99035231 on Build GRCh38
rs114250093 Chr.2: 99035138-99035138 on Build GRCh38
rs976640995 Chr.2: 99035121-99035121 on Build GRCh38
rs776204110 Chr.2: 99035201-99035201 on Build GRCh38
rs764917423 Chr.2: 99035219-99035219 on Build GRCh38
rs200196078 Chr.2: 99035264-99035264 on Build GRCh38
rs747012861 Chr.2: 99035275-99035275 on Build GRCh38
rs147204896 Chr.2: 99035271-99035271 on Build GRCh38
rs756624311 Chr.2: 99035238-99035238 on Build GRCh38
rs951231884 Chr.2: 99035144-99035144 on Build GRCh38
rs995918466 Chr.2: 99035107-99035107 on Build GRCh38
rs758201155 Chr.2: 99035267-99035267 on Build GRCh38
rs755195469 Chr.2: 99035244-99035244 on Build GRCh38
rs776057459 Chr.2: 99035293-99035293 on Build GRCh38
rs746587773 Chr.2: 99035183-99035184 on Build GRCh38
rs770837918 Chr.2: 99035287-99035287 on Build GRCh38
rs759164735 Chr.2: 99035207-99035207 on Build GRCh38
rs1020635722 Chr.2: 99035209-99035209 on Build GRCh38
rs989977907 Chr.2: 99035156-99035156 on Build GRCh38
rs781692720 Chr.2: 99035225-99035225 on Build GRCh38
rs116333015 Chr.2: 99035220-99035220 on Build GRCh38
rs779225163 Chr.2: 99035253-99035253 on Build GRCh38
rs909649888 Chr.2: 99035176-99035176 on Build GRCh38
rs967257497 Chr.2: 99035213-99035213 on Build GRCh38
rs766435883 Chr.2: 99035239-99035239 on Build GRCh38
rs750982829 Chr.2: 99035235-99035235 on Build GRCh38
rs753931998 Chr.2: 99035241-99035241 on Build GRCh38
rs915581527 Chr.2: 99035232-99035232 on Build GRCh38
rs771404043 Chr.2: 99035184-99035184 on Build GRCh38
rs928628984 Chr.2: 99035229-99035229 on Build GRCh38
rs982542605 Chr.2: 99035162-99035162 on Build GRCh38

View Full Product Details