Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 101164278-101164523 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
VGF
Entrez Gene ID
7425
Gene Name
VGF nerve growth factor inducible
Gene Aliases
SCG7, SgVII
UniGene
Hs.587325
Gene Chromosome Location
Chr.7: 101162509-101169956 on Build GRCh38
Transcripts
NM_003378.3 XM_005250561.4 XM_011516549.2 XM_017012579.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3876626 c.520G>A p.A174T Chr.7: 101164324-101164324 on Build GRCh38
398334 c.394C>A p.P132T Chr.7: 101164450-101164450 on Build GRCh38
6247224 c.490C>A p.Q164K Chr.7: 101164354-101164354 on Build GRCh38
3631327 c.540G>C p.T180T Chr.7: 101164304-101164304 on Build GRCh38
6398367 c.351G>T p.E117D Chr.7: 101164493-101164493 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768425527 Chr.7: 101164477-101164477 on Build GRCh38
rs765733381 Chr.7: 101164304-101164304 on Build GRCh38
rs946129418 Chr.7: 101164468-101164468 on Build GRCh38
rs759028390 Chr.7: 101164326-101164326 on Build GRCh38
rs775158895 Chr.7: 101164295-101164295 on Build GRCh38
rs989757965 Chr.7: 101164441-101164441 on Build GRCh38
rs762298837 Chr.7: 101164302-101164302 on Build GRCh38
rs778745394 Chr.7: 101164445-101164445 on Build GRCh38
rs745858208 Chr.7: 101164469-101164469 on Build GRCh38
rs766812402 Chr.7: 101164399-101164399 on Build GRCh38
rs777969426 Chr.7: 101164418-101164418 on Build GRCh38
rs372301855 Chr.7: 101164405-101164405 on Build GRCh38
rs1022284633 Chr.7: 101164474-101164474 on Build GRCh38
rs763436638 Chr.7: 101164495-101164495 on Build GRCh38
rs755030999 Chr.7: 101164342-101164342 on Build GRCh38
rs751597749 Chr.7: 101164333-101164333 on Build GRCh38
rs980676864 Chr.7: 101164356-101164356 on Build GRCh38
rs535664776 Chr.7: 101164439-101164439 on Build GRCh38
rs773777980 Chr.7: 101164394-101164394 on Build GRCh38
rs763822742 Chr.7: 101164389-101164389 on Build GRCh38
rs956954541 Chr.7: 101164424-101164424 on Build GRCh38
rs774284715 Chr.7: 101164370-101164370 on Build GRCh38
rs749093213 Chr.7: 101164433-101164433 on Build GRCh38
rs776367894 Chr.7: 101164483-101164483 on Build GRCh38
rs756387061 Chr.7: 101164415-101164415 on Build GRCh38
rs958078586 Chr.7: 101164360-101164360 on Build GRCh38
rs766902706 Chr.7: 101164327-101164327 on Build GRCh38
rs750791722 Chr.7: 101164320-101164320 on Build GRCh38
rs774828122 Chr.7: 101164373-101164373 on Build GRCh38
rs745390302 Chr.7: 101164371-101164371 on Build GRCh38
rs781296347 Chr.7: 101164345-101164345 on Build GRCh38
rs376280558 Chr.7: 101164338-101164338 on Build GRCh38
rs777594782 Chr.7: 101164358-101164358 on Build GRCh38
rs752060603 Chr.7: 101164402-101164402 on Build GRCh38
rs1041727282 Chr.7: 101164500-101164500 on Build GRCh38
rs748333973 Chr.7: 101164346-101164346 on Build GRCh38
rs756298712 Chr.7: 101164351-101164351 on Build GRCh38
rs771878639 Chr.7: 101164475-101164475 on Build GRCh38
rs749070087 Chr.7: 101164365-101164365 on Build GRCh38
rs557175283 Chr.7: 101164476-101164476 on Build GRCh38
rs770923806 Chr.7: 101164367-101164367 on Build GRCh38
rs1024609554 Chr.7: 101164328-101164328 on Build GRCh38
rs750986094 Chr.7: 101164472-101164474 on Build GRCh38
rs199908448 Chr.7: 101164409-101164409 on Build GRCh38
rs760450108 Chr.7: 101164382-101164382 on Build GRCh38

View Full Product Details