Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 62003290-62003555 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PTPRG
Entrez Gene ID
5793
Gene Name
protein tyrosine phosphatase, receptor type G
Gene Aliases
HPTPG, PTPG, R-PTP-GAMMA, RPTPG
UniGene
Hs.595541
Gene Chromosome Location
Chr.3: 61561569-62297609 on Build GRCh38
Transcripts
NM_002841.3 XM_005265353.3 XM_017006961.1 XM_017006962.1 XM_017006963.1 XM_017006964.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1579531 c.478G>T p.G160C Chr.3: 62003456-62003456 on Build GRCh38
227717 c.455G>T p.W152L Chr.3: 62003433-62003433 on Build GRCh38
1617980 c.470G>T p.G157V Chr.3: 62003448-62003448 on Build GRCh38
1309421 c.390C>T p.D130D Chr.3: 62003368-62003368 on Build GRCh38
1309422 c.433G>T p.A145S Chr.3: 62003411-62003411 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748575600 Chr.3: 62003520-62003520 on Build GRCh38
rs140851159 Chr.3: 62003422-62003422 on Build GRCh38
rs750068236 Chr.3: 62003394-62003394 on Build GRCh38
rs769062369 Chr.3: 62003346-62003346 on Build GRCh38
rs189356599 Chr.3: 62003368-62003368 on Build GRCh38
rs770279523 Chr.3: 62003326-62003326 on Build GRCh38
rs766805370 Chr.3: 62003397-62003398 on Build GRCh38
rs775226099 Chr.3: 62003452-62003452 on Build GRCh38
rs1038831841 Chr.3: 62003458-62003458 on Build GRCh38
rs755442122 Chr.3: 62003420-62003420 on Build GRCh38
rs138601102 Chr.3: 62003522-62003522 on Build GRCh38
rs2240274 Chr.3: 62003334-62003334 on Build GRCh38
rs150122281 Chr.3: 62003518-62003518 on Build GRCh38
rs752539487 Chr.3: 62003369-62003369 on Build GRCh38
rs201734186 Chr.3: 62003445-62003445 on Build GRCh38
rs374973218 Chr.3: 62003471-62003471 on Build GRCh38
rs368752966 Chr.3: 62003319-62003319 on Build GRCh38
rs140777402 Chr.3: 62003354-62003354 on Build GRCh38
rs774150743 Chr.3: 62003469-62003469 on Build GRCh38
rs749788211 Chr.3: 62003345-62003345 on Build GRCh38
rs749642322 Chr.3: 62003411-62003411 on Build GRCh38
rs758486142 Chr.3: 62003331-62003333 on Build GRCh38
rs373590197 Chr.3: 62003373-62003373 on Build GRCh38
rs766130550 Chr.3: 62003486-62003486 on Build GRCh38
rs755784901 Chr.3: 62003483-62003483 on Build GRCh38
rs755877507 Chr.3: 62003397-62003397 on Build GRCh38
rs370339396 Chr.3: 62003370-62003370 on Build GRCh38
rs908850824 Chr.3: 62003416-62003416 on Build GRCh38
rs748752255 Chr.3: 62003424-62003424 on Build GRCh38
rs758480466 Chr.3: 62003453-62003453 on Build GRCh38
rs745574653 Chr.3: 62003322-62003322 on Build GRCh38
rs371488686 Chr.3: 62003455-62003455 on Build GRCh38
rs780914955 Chr.3: 62003391-62003391 on Build GRCh38
rs773413749 Chr.3: 62003352-62003352 on Build GRCh38
rs761799368 Chr.3: 62003350-62003350 on Build GRCh38
rs367774512 Chr.3: 62003454-62003454 on Build GRCh38
rs755287937 Chr.3: 62003516-62003516 on Build GRCh38
rs553923548 Chr.3: 62003438-62003438 on Build GRCh38
rs747211567 Chr.3: 62003442-62003442 on Build GRCh38
rs761849753 Chr.3: 62003470-62003470 on Build GRCh38
rs867441157 Chr.3: 62003437-62003437 on Build GRCh38
rs757115800 Chr.3: 62003448-62003448 on Build GRCh38
rs772283947 Chr.3: 62003351-62003351 on Build GRCh38
rs772137991 Chr.3: 62003435-62003435 on Build GRCh38
rs780514305 Chr.3: 62003408-62003408 on Build GRCh38
rs372067576 Chr.3: 62003475-62003475 on Build GRCh38
rs771166810 Chr.3: 62003450-62003450 on Build GRCh38
rs756893775 Chr.3: 62003318-62003318 on Build GRCh38
rs754169873 Chr.3: 62003493-62003493 on Build GRCh38
rs933147386 Chr.3: 62003510-62003510 on Build GRCh38
rs777783036 Chr.3: 62003521-62003521 on Build GRCh38
rs780608111 Chr.3: 62003342-62003342 on Build GRCh38

View Full Product Details