Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 15949767-15949992 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCC1
Entrez Gene ID
4363
Gene Name
ATP binding cassette subfamily C member 1
Gene Aliases
ABC29, ABCC, GS-X, MRP, MRP1
UniGene
Hs.391464
Gene Chromosome Location
Chr.16: 15949577-16143074 on Build GRCh38
Transcripts
NM_004996.3

SNP Detail

SNP ID Chromosome Location
rs900595137 Chr.16: 15949959-15949959 on Build GRCh38
rs1056102741 Chr.16: 15949853-15949853 on Build GRCh38
rs996249913 Chr.16: 15949961-15949961 on Build GRCh38
rs972255080 Chr.16: 15949815-15949815 on Build GRCh38
rs939100964 Chr.16: 15949832-15949832 on Build GRCh38
rs936482010 Chr.16: 15949939-15949939 on Build GRCh38
rs865996004 Chr.16: 15949893-15949893 on Build GRCh38
rs909494728 Chr.16: 15949794-15949794 on Build GRCh38
rs1043644413 Chr.16: 15949902-15949902 on Build GRCh38
rs768460096 Chr.16: 15949941-15949941 on Build GRCh38
rs1002822908 Chr.16: 15949852-15949852 on Build GRCh38
rs560199163 Chr.16: 15949969-15949969 on Build GRCh38
rs918311722 Chr.16: 15949818-15949818 on Build GRCh38
rs533486651 Chr.16: 15949906-15949906 on Build GRCh38
rs182752194 Chr.16: 15949804-15949804 on Build GRCh38
rs999350755 Chr.16: 15949908-15949908 on Build GRCh38
rs562494702 Chr.16: 15949784-15949784 on Build GRCh38
rs372392599 Chr.16: 15949910-15949910 on Build GRCh38
rs889120436 Chr.16: 15949964-15949964 on Build GRCh38
rs1053902242 Chr.16: 15949949-15949949 on Build GRCh38
rs997566879 Chr.16: 15949958-15949958 on Build GRCh38
rs1052240859 Chr.16: 15949930-15949930 on Build GRCh38
rs1051772392 Chr.16: 15949967-15949967 on Build GRCh38
rs916257189 Chr.16: 15949886-15949892 on Build GRCh38
rs953725451 Chr.16: 15949783-15949783 on Build GRCh38
rs901898913 Chr.16: 15949946-15949946 on Build GRCh38

View Full Product Details