Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 44118333-44118604 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF225
Entrez Gene ID
7768
Gene Name
zinc finger protein 225
Gene Aliases
-
UniGene
Hs.279567
Gene Chromosome Location
Chr.19: 44111426-44134816 on Build GRCh38
Transcripts
NM_001321685.1 NM_013362.3 XM_011527285.2 XM_011527286.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6017536 c.183G>C p.K61N Chr.19: 44118522-44118522 on Build GRCh38
997819 c.165T>C p.D55D Chr.19: 44118504-44118504 on Build GRCh38
1168876 c.178C>T p.L60L Chr.19: 44118517-44118517 on Build GRCh38
5984684 c.205G>C p.E69Q Chr.19: 44118544-44118544 on Build GRCh38
5555611 c.144G>A p.G48G Chr.19: 44118483-44118483 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs370346059 Chr.19: 44118454-44118454 on Build GRCh38
rs773921352 Chr.19: 44118519-44118519 on Build GRCh38
rs868601142 Chr.19: 44118403-44118403 on Build GRCh38
rs944469371 Chr.19: 44118483-44118483 on Build GRCh38
rs745804099 Chr.19: 44118485-44118485 on Build GRCh38
rs757201822 Chr.19: 44118479-44118479 on Build GRCh38
rs748227060 Chr.19: 44118542-44118543 on Build GRCh38
rs540946151 Chr.19: 44118480-44118480 on Build GRCh38
rs762208576 Chr.19: 44118507-44118507 on Build GRCh38
rs1031298277 Chr.19: 44118491-44118491 on Build GRCh38
rs371799405 Chr.19: 44118578-44118578 on Build GRCh38
rs906634382 Chr.19: 44118361-44118361 on Build GRCh38
rs752903565 Chr.19: 44118572-44118572 on Build GRCh38
rs765768361 Chr.19: 44118437-44118437 on Build GRCh38
rs558322207 Chr.19: 44118395-44118395 on Build GRCh38
rs751544140 Chr.19: 44118554-44118554 on Build GRCh38
rs747492231 Chr.19: 44118504-44118504 on Build GRCh38
rs1051886406 Chr.19: 44118406-44118407 on Build GRCh38
rs751846827 Chr.19: 44118473-44118473 on Build GRCh38
rs940180003 Chr.19: 44118522-44118522 on Build GRCh38
rs758561332 Chr.19: 44118461-44118461 on Build GRCh38
rs76696522 Chr.19: 44118527-44118527 on Build GRCh38
rs989450031 Chr.19: 44118404-44118404 on Build GRCh38
rs766362247 Chr.19: 44118462-44118462 on Build GRCh38
rs974374861 Chr.19: 44118486-44118486 on Build GRCh38
rs772503035 Chr.19: 44118549-44118550 on Build GRCh38
rs755213333 Chr.19: 44118473-44118473 on Build GRCh38
rs373963033 Chr.19: 44118474-44118474 on Build GRCh38
rs1003505968 Chr.19: 44118367-44118367 on Build GRCh38
rs888708572 Chr.19: 44118517-44118517 on Build GRCh38
rs767670071 Chr.19: 44118569-44118569 on Build GRCh38
rs768150427 Chr.19: 44118495-44118495 on Build GRCh38
rs34863330 Chr.19: 44118488-44118488 on Build GRCh38
rs778925210 Chr.19: 44118516-44118516 on Build GRCh38
rs114271963 Chr.19: 44118489-44118489 on Build GRCh38
rs895745901 Chr.19: 44118499-44118499 on Build GRCh38
rs770267059 Chr.19: 44118514-44118514 on Build GRCh38
rs191225974 Chr.19: 44118476-44118476 on Build GRCh38
rs764549796 Chr.19: 44118550-44118551 on Build GRCh38
rs748611472 Chr.19: 44118396-44118396 on Build GRCh38
rs746700398 Chr.19: 44118493-44118493 on Build GRCh38
rs1002319676 Chr.19: 44118538-44118538 on Build GRCh38
rs563327487 Chr.19: 44118545-44118545 on Build GRCh38
rs762437672 Chr.19: 44118360-44118360 on Build GRCh38
rs777091355 Chr.19: 44118356-44118356 on Build GRCh38
rs759809853 Chr.19: 44118557-44118557 on Build GRCh38

View Full Product Details