Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 62578883-62579110 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TUT1
Entrez Gene ID
64852
Gene Name
terminal uridylyl transferase 1, U6 snRNA-specific
Gene Aliases
PAPD2, RBM21, STARPAP, TUTase, URLC6
UniGene
Hs.728983
Gene Chromosome Location
Chr.11: 62575045-62591637 on Build GRCh38
Transcripts
NM_022830.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3986303 c.788A>G p.D263G Chr.11: 62578933-62578933 on Build GRCh38
7259400 c.917C>T p.A306V Chr.11: 62578918-62578918 on Build GRCh38
5970245 c.809T>C p.V270A Chr.11: 62579026-62579026 on Build GRCh38
5970246 c.695T>C p.V232A Chr.11: 62579026-62579026 on Build GRCh38
7281427 c.897T>C p.P299P Chr.11: 62578938-62578938 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374660195 Chr.11: 62578997-62578997 on Build GRCh38
rs116766958 Chr.11: 62579038-62579038 on Build GRCh38
rs908251878 Chr.11: 62578908-62578908 on Build GRCh38
rs144066512 Chr.11: 62579032-62579033 on Build GRCh38
rs771430657 Chr.11: 62578976-62578976 on Build GRCh38
rs199576589 Chr.11: 62578948-62578948 on Build GRCh38
rs767277277 Chr.11: 62578996-62578996 on Build GRCh38
rs760625974 Chr.11: 62579001-62579001 on Build GRCh38
rs747386899 Chr.11: 62579050-62579050 on Build GRCh38
rs376663677 Chr.11: 62578919-62578919 on Build GRCh38
rs773911517 Chr.11: 62578989-62578989 on Build GRCh38
rs761643438 Chr.11: 62578992-62578992 on Build GRCh38
rs1010499230 Chr.11: 62579031-62579031 on Build GRCh38
rs748505320 Chr.11: 62578959-62578959 on Build GRCh38
rs757709024 Chr.11: 62579060-62579060 on Build GRCh38
rs1007429721 Chr.11: 62578934-62578934 on Build GRCh38
rs747535539 Chr.11: 62578965-62578965 on Build GRCh38
rs748816245 Chr.11: 62578981-62578981 on Build GRCh38
rs758784569 Chr.11: 62579033-62579033 on Build GRCh38
rs765468114 Chr.11: 62579023-62579023 on Build GRCh38
rs755194402 Chr.11: 62578937-62578937 on Build GRCh38
rs755141254 Chr.11: 62579017-62579017 on Build GRCh38
rs1038617940 Chr.11: 62578918-62578918 on Build GRCh38
rs774921483 Chr.11: 62578979-62578979 on Build GRCh38
rs544541221 Chr.11: 62578961-62578961 on Build GRCh38
rs893753353 Chr.11: 62578969-62578969 on Build GRCh38
rs1021046033 Chr.11: 62579052-62579052 on Build GRCh38
rs368086926 Chr.11: 62579071-62579071 on Build GRCh38
rs753922561 Chr.11: 62579002-62579002 on Build GRCh38
rs775403051 Chr.11: 62579048-62579048 on Build GRCh38
rs555954379 Chr.11: 62578909-62578909 on Build GRCh38
rs903877989 Chr.11: 62579054-62579054 on Build GRCh38
rs770382959 Chr.11: 62579032-62579032 on Build GRCh38
rs768255294 Chr.11: 62578985-62578985 on Build GRCh38
rs766635480 Chr.11: 62578911-62578911 on Build GRCh38
rs1020595546 Chr.11: 62579035-62579035 on Build GRCh38

View Full Product Details