Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 216112941-216113202 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
XRCC5
Entrez Gene ID
7520
Gene Name
X-ray repair cross complementing 5
Gene Aliases
KARP-1, KARP1, KU80, KUB2, Ku86, NFIV
UniGene
Hs.388739
Gene Chromosome Location
Chr.2: 216109297-216206293 on Build GRCh38
Transcripts
NM_021141.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6494785 c.124G>A p.V42I Chr.2: 216113118-216113118 on Build GRCh38
1691983 c.88_89CC>TT p.P30L Chr.2: 216113082-216113083 on Build GRCh38
1691982 c.86C>T p.S29F Chr.2: 216113080-216113080 on Build GRCh38
6446586 c.127C>T p.Q43* Chr.2: 216113121-216113121 on Build GRCh38
6888098 c.135G>A p.Q45Q Chr.2: 216113129-216113129 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs377730441 Chr.2: 216113163-216113163 on Build GRCh38
rs757885210 Chr.2: 216113067-216113067 on Build GRCh38
rs958827020 Chr.2: 216113164-216113164 on Build GRCh38
rs1020875534 Chr.2: 216113102-216113102 on Build GRCh38
rs769403229 Chr.2: 216113154-216113154 on Build GRCh38
rs771918326 Chr.2: 216112979-216112979 on Build GRCh38
rs764581950 Chr.2: 216113051-216113051 on Build GRCh38
rs745746809 Chr.2: 216113142-216113142 on Build GRCh38
rs749582080 Chr.2: 216113108-216113108 on Build GRCh38
rs776552591 Chr.2: 216113140-216113140 on Build GRCh38
rs182171056 Chr.2: 216113006-216113006 on Build GRCh38
rs746786664 Chr.2: 216113137-216113137 on Build GRCh38
rs754627317 Chr.2: 216112979-216112980 on Build GRCh38
rs769459117 Chr.2: 216113071-216113071 on Build GRCh38
rs770825706 Chr.2: 216112996-216112996 on Build GRCh38
rs1051169521 Chr.2: 216113034-216113034 on Build GRCh38
rs373315231 Chr.2: 216113139-216113139 on Build GRCh38
rs41296930 Chr.2: 216112978-216112978 on Build GRCh38
rs55662197 Chr.2: 216112976-216112976 on Build GRCh38
rs768163718 Chr.2: 216113074-216113074 on Build GRCh38
rs755152526 Chr.2: 216113118-216113118 on Build GRCh38
rs769816648 Chr.2: 216113039-216113039 on Build GRCh38
rs56170971 Chr.2: 216113004-216113004 on Build GRCh38
rs377451599 Chr.2: 216113175-216113175 on Build GRCh38
rs149406399 Chr.2: 216113160-216113160 on Build GRCh38
rs756226383 Chr.2: 216113099-216113099 on Build GRCh38
rs80130892 Chr.2: 216112975-216112975 on Build GRCh38
rs750922846 Chr.2: 216113093-216113093 on Build GRCh38
rs779944364 Chr.2: 216113103-216113103 on Build GRCh38
rs889241859 Chr.2: 216112990-216112990 on Build GRCh38
rs760673986 Chr.2: 216112988-216112988 on Build GRCh38
rs979478586 Chr.2: 216113162-216113162 on Build GRCh38
rs780756006 Chr.2: 216113165-216113166 on Build GRCh38
rs374905283 Chr.2: 216113174-216113174 on Build GRCh38
rs774096336 Chr.2: 216113178-216113178 on Build GRCh38
rs764584130 Chr.2: 216113009-216113009 on Build GRCh38
rs772869384 Chr.2: 216112982-216112982 on Build GRCh38
rs751938865 Chr.2: 216113064-216113064 on Build GRCh38
rs777586585 Chr.2: 216113122-216113122 on Build GRCh38
rs775220412 Chr.2: 216113025-216113025 on Build GRCh38

View Full Product Details