Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 488808-489059 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RAB11FIP3
Entrez Gene ID
9727
Gene Name
RAB11 family interacting protein 3
Gene Aliases
CART1, Rab11-FIP3
UniGene
Hs.531642
Gene Chromosome Location
Chr.16: 425621-522485 on Build GRCh38
Transcripts
NM_001142272.1 NM_014700.3 XM_005255713.3 XM_005255714.1 XM_005255715.4 XM_005255717.3 XM_005255718.3 XM_011522764.2 XM_011522765.2 XM_017023907.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4883797 c.1265G>T p.S422I Chr.16: 489000-489000 on Build GRCh38
4061227 c.1265+1G>T p.? Chr.16: 489001-489001 on Build GRCh38
5916541 c.1136C>T p.S379F Chr.16: 488871-488871 on Build GRCh38
6496892 c.278A>G p.H93R Chr.16: 488901-488901 on Build GRCh38
4260633 c.1249G>A p.A417T Chr.16: 488984-488984 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs558144377 Chr.16: 488932-488932 on Build GRCh38
rs763319081 Chr.16: 488886-488886 on Build GRCh38
rs752519617 Chr.16: 488940-488940 on Build GRCh38
rs143244233 Chr.16: 488993-488993 on Build GRCh38
rs758328581 Chr.16: 488947-488947 on Build GRCh38
rs139898544 Chr.16: 488888-488888 on Build GRCh38
rs140423430 Chr.16: 488969-488969 on Build GRCh38
rs762022224 Chr.16: 489032-489032 on Build GRCh38
rs1007131348 Chr.16: 488859-488859 on Build GRCh38
rs758421303 Chr.16: 489008-489008 on Build GRCh38
rs554359406 Chr.16: 488863-488863 on Build GRCh38
rs150937967 Chr.16: 488995-488995 on Build GRCh38
rs777712988 Chr.16: 488955-488955 on Build GRCh38
rs369419129 Chr.16: 488927-488927 on Build GRCh38
rs746028307 Chr.16: 488959-488959 on Build GRCh38
rs536304412 Chr.16: 488862-488862 on Build GRCh38
rs748505585 Chr.16: 488984-488984 on Build GRCh38
rs756259546 Chr.16: 489025-489025 on Build GRCh38
rs762211544 Chr.16: 488894-488894 on Build GRCh38
rs756379921 Chr.16: 488960-488960 on Build GRCh38
rs760328450 Chr.16: 488923-488923 on Build GRCh38
rs745932166 Chr.16: 488880-488880 on Build GRCh38
rs35538165 Chr.16: 488938-488938 on Build GRCh38
rs749422181 Chr.16: 488965-488965 on Build GRCh38
rs372735637 Chr.16: 488931-488931 on Build GRCh38
rs778528267 Chr.16: 488846-488846 on Build GRCh38
rs202139275 Chr.16: 489009-489009 on Build GRCh38
rs146483534 Chr.16: 488881-488881 on Build GRCh38
rs755149756 Chr.16: 489036-489036 on Build GRCh38
rs780368238 Chr.16: 489027-489027 on Build GRCh38
rs377509491 Chr.16: 488994-488994 on Build GRCh38
rs960523827 Chr.16: 488976-488976 on Build GRCh38
rs754959809 Chr.16: 488845-488845 on Build GRCh38
rs772358681 Chr.16: 488986-488986 on Build GRCh38
rs779956841 Chr.16: 488964-488964 on Build GRCh38
rs753654226 Chr.16: 488836-488836 on Build GRCh38
rs760275499 Chr.16: 488988-488988 on Build GRCh38
rs149810973 Chr.16: 488855-488855 on Build GRCh38
rs773583446 Chr.16: 488987-488987 on Build GRCh38
rs749311559 Chr.16: 489033-489033 on Build GRCh38
rs538129861 Chr.16: 488914-488914 on Build GRCh38
rs764695211 Chr.16: 488861-488861 on Build GRCh38
rs978944688 Chr.16: 488951-488951 on Build GRCh38
rs375253048 Chr.16: 489012-489012 on Build GRCh38
rs752750674 Chr.16: 488996-488996 on Build GRCh38
rs774892939 Chr.16: 488983-488983 on Build GRCh38
rs570846363 Chr.16: 488847-488847 on Build GRCh38
rs767140800 Chr.16: 488899-488899 on Build GRCh38
rs747987695 Chr.16: 488860-488860 on Build GRCh38
rs911655984 Chr.16: 488985-488985 on Build GRCh38
rs1034817684 Chr.16: 488887-488887 on Build GRCh38
rs375661104 Chr.16: 488858-488858 on Build GRCh38
rs139836747 Chr.16: 488893-488893 on Build GRCh38

View Full Product Details