Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 67911213-67911486 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMPRSS11A
Entrez Gene ID
339967
Gene Name
transmembrane protease, serine 11A
Gene Aliases
ECRG1
UniGene
Hs.450110
Gene Chromosome Location
Chr.4: 67910301-67963514 on Build GRCh38
Transcripts
NM_001114387.1 NM_182606.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5750886 c.1169T>C p.V390A Chr.4: 67911439-67911439 on Build GRCh38
2828931 c.1153T>C p.Y385H Chr.4: 67911455-67911455 on Build GRCh38
2828930 c.1250C>A p.S417* Chr.4: 67911358-67911358 on Build GRCh38
4841159 c.1266A>C p.*422Y Chr.4: 67911342-67911342 on Build GRCh38
3826084 c.1178G>A p.G393E Chr.4: 67911430-67911430 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776762233 Chr.4: 67911370-67911370 on Build GRCh38
rs548530297 Chr.4: 67911306-67911306 on Build GRCh38
rs774683038 Chr.4: 67911300-67911300 on Build GRCh38
rs746998961 Chr.4: 67911428-67911428 on Build GRCh38
rs749597665 Chr.4: 67911296-67911296 on Build GRCh38
rs371262351 Chr.4: 67911337-67911337 on Build GRCh38
rs1004143120 Chr.4: 67911349-67911349 on Build GRCh38
rs201118203 Chr.4: 67911439-67911439 on Build GRCh38
rs768874044 Chr.4: 67911382-67911382 on Build GRCh38
rs944094312 Chr.4: 67911284-67911284 on Build GRCh38
rs760403424 Chr.4: 67911316-67911316 on Build GRCh38
rs769088421 Chr.4: 67911298-67911298 on Build GRCh38
rs28648375 Chr.4: 67911282-67911282 on Build GRCh38
rs895485673 Chr.4: 67911378-67911378 on Build GRCh38
rs12646286 Chr.4: 67911336-67911336 on Build GRCh38
rs114594588 Chr.4: 67911323-67911323 on Build GRCh38
rs368829002 Chr.4: 67911304-67911304 on Build GRCh38
rs753551188 Chr.4: 67911329-67911329 on Build GRCh38
rs762794520 Chr.4: 67911459-67911459 on Build GRCh38
rs148638761 Chr.4: 67911456-67911456 on Build GRCh38
rs750435881 Chr.4: 67911346-67911346 on Build GRCh38
rs1056765315 Chr.4: 67911340-67911340 on Build GRCh38
rs375027937 Chr.4: 67911345-67911345 on Build GRCh38
rs1038833130 Chr.4: 67911288-67911288 on Build GRCh38
rs376494815 Chr.4: 67911411-67911411 on Build GRCh38
rs369203246 Chr.4: 67911374-67911374 on Build GRCh38
rs373272353 Chr.4: 67911387-67911387 on Build GRCh38
rs887223448 Chr.4: 67911348-67911348 on Build GRCh38
rs765485351 Chr.4: 67911429-67911430 on Build GRCh38
rs780260050 Chr.4: 67911291-67911291 on Build GRCh38
rs748593910 Chr.4: 67911389-67911389 on Build GRCh38
rs770583995 Chr.4: 67911432-67911432 on Build GRCh38
rs760201060 Chr.4: 67911420-67911420 on Build GRCh38
rs756200756 Chr.4: 67911366-67911366 on Build GRCh38
rs762005747 Chr.4: 67911410-67911410 on Build GRCh38
rs758409731 Chr.4: 67911343-67911343 on Build GRCh38
rs914467913 Chr.4: 67911272-67911272 on Build GRCh38
rs772919156 Chr.4: 67911315-67911315 on Build GRCh38
rs773855875 Chr.4: 67911416-67911416 on Build GRCh38
rs562773026 Chr.4: 67911373-67911373 on Build GRCh38
rs939680050 Chr.4: 67911339-67911339 on Build GRCh38
rs759271084 Chr.4: 67911446-67911446 on Build GRCh38

View Full Product Details