Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 14256644-14256869 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SPON1
Entrez Gene ID
10418
Gene Name
spondin 1
Gene Aliases
VSGP/F-spondin, f-spondin
UniGene
Hs.643864
Gene Chromosome Location
Chr.11: 13962637-14268133 on Build GRCh38
Transcripts
NM_006108.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6789204 c.1306+4_1306+5insG p.? Chr.11: 14256696-14256697 on Build GRCh38
4447217 c.1300G>A p.D434N Chr.11: 14256686-14256686 on Build GRCh38
6789196 c.1301A>T p.D434V Chr.11: 14256687-14256687 on Build GRCh38
3666881 c.1290A>C p.P430P Chr.11: 14256676-14256676 on Build GRCh38
6249243 c.1306+5G>T p.? Chr.11: 14256697-14256697 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782111883 Chr.11: 14256676-14256676 on Build GRCh38
rs371965820 Chr.11: 14256686-14256686 on Build GRCh38
rs369824152 Chr.11: 14256697-14256697 on Build GRCh38
rs782549194 Chr.11: 14256815-14256815 on Build GRCh38
rs782230767 Chr.11: 14256739-14256739 on Build GRCh38
rs567781903 Chr.11: 14256847-14256847 on Build GRCh38
rs374070123 Chr.11: 14256712-14256712 on Build GRCh38
rs781997044 Chr.11: 14256674-14256674 on Build GRCh38
rs782565447 Chr.11: 14256692-14256692 on Build GRCh38
rs782628034 Chr.11: 14256731-14256731 on Build GRCh38
rs376572740 Chr.11: 14256687-14256687 on Build GRCh38
rs555976662 Chr.11: 14256817-14256817 on Build GRCh38
rs782532257 Chr.11: 14256728-14256728 on Build GRCh38
rs933842139 Chr.11: 14256779-14256779 on Build GRCh38
rs781889696 Chr.11: 14256726-14256726 on Build GRCh38
rs782445397 Chr.11: 14256715-14256715 on Build GRCh38
rs374898036 Chr.11: 14256725-14256725 on Build GRCh38
rs941255637 Chr.11: 14256696-14256696 on Build GRCh38

View Full Product Details