Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 30187051-30187278 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CORO1A
Entrez Gene ID
11151
Gene Name
coronin 1A
Gene Aliases
CLABP, CLIPINA, HCORO1, IMD8, TACO, p57
UniGene
Hs.415067
Gene Chromosome Location
Chr.16: 30183410-30189076 on Build GRCh38
Transcripts
NM_001193333.2 NM_007074.3 XM_011545714.2 XM_017022885.1 XM_017022886.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5517210 c.487A>G p.T163A Chr.16: 30187074-30187074 on Build GRCh38
1678996 c.602G>A p.R201H Chr.16: 30187189-30187189 on Build GRCh38
5485789 c.495G>A p.A165A Chr.16: 30187082-30187082 on Build GRCh38
4674221 c.475T>C p.W159R Chr.16: 30187062-30187062 on Build GRCh38
6634193 c.536T>C p.I179T Chr.16: 30187123-30187123 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs531482387 Chr.16: 30187184-30187184 on Build GRCh38
rs141746241 Chr.16: 30187148-30187148 on Build GRCh38
rs756032147 Chr.16: 30187200-30187200 on Build GRCh38
rs765799047 Chr.16: 30187167-30187167 on Build GRCh38
rs752499984 Chr.16: 30187197-30187197 on Build GRCh38
rs551747429 Chr.16: 30187120-30187120 on Build GRCh38
rs751894231 Chr.16: 30187185-30187185 on Build GRCh38
rs748496776 Chr.16: 30187252-30187252 on Build GRCh38
rs570148791 Chr.16: 30187121-30187121 on Build GRCh38
rs931113520 Chr.16: 30187146-30187146 on Build GRCh38
rs767224016 Chr.16: 30187196-30187196 on Build GRCh38
rs548533633 Chr.16: 30187183-30187183 on Build GRCh38
rs370019525 Chr.16: 30187246-30187246 on Build GRCh38
rs371818902 Chr.16: 30187218-30187218 on Build GRCh38
rs548894681 Chr.16: 30187152-30187152 on Build GRCh38
rs369595327 Chr.16: 30187127-30187127 on Build GRCh38
rs148312916 Chr.16: 30187144-30187144 on Build GRCh38
rs760274904 Chr.16: 30187230-30187230 on Build GRCh38
rs199752871 Chr.16: 30187106-30187106 on Build GRCh38
rs755211107 Chr.16: 30187188-30187188 on Build GRCh38
rs375950115 Chr.16: 30187231-30187231 on Build GRCh38
rs527428900 Chr.16: 30187087-30187087 on Build GRCh38
rs763170903 Chr.16: 30187182-30187182 on Build GRCh38
rs769707848 Chr.16: 30187107-30187107 on Build GRCh38
rs145989561 Chr.16: 30187191-30187191 on Build GRCh38
rs371653086 Chr.16: 30187081-30187081 on Build GRCh38
rs762228775 Chr.16: 30187164-30187164 on Build GRCh38
rs746348200 Chr.16: 30187208-30187209 on Build GRCh38
rs139989282 Chr.16: 30187217-30187217 on Build GRCh38
rs777740076 Chr.16: 30187204-30187204 on Build GRCh38
rs1046871825 Chr.16: 30187165-30187165 on Build GRCh38
rs374849081 Chr.16: 30187082-30187082 on Build GRCh38
rs771687134 Chr.16: 30187239-30187239 on Build GRCh38
rs748052416 Chr.16: 30187093-30187093 on Build GRCh38
rs780371160 Chr.16: 30187077-30187077 on Build GRCh38
rs745531321 Chr.16: 30187142-30187142 on Build GRCh38

View Full Product Details