Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 30927406-30927667 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXL19
Entrez Gene ID
54620
Gene Name
F-box and leucine rich repeat protein 19
Gene Aliases
CXXC11, Fbl19, JHDM1C
UniGene
Hs.152149
Gene Chromosome Location
Chr.16: 30923055-30948783 on Build GRCh38
Transcripts
NM_001099784.2 NM_001282351.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5850762 c.341G>A p.C114Y Chr.16: 30927411-30927411 on Build GRCh38
6555632 c.447C>T p.T149T Chr.16: 30927638-30927638 on Build GRCh38
6248371 c.355G>T p.E119* Chr.16: 30927425-30927425 on Build GRCh38
5731324 c.410C>T p.A137V Chr.16: 30927601-30927601 on Build GRCh38
5960022 c.431A>T p.E144V Chr.16: 30927622-30927622 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs12930545 Chr.16: 30927490-30927490 on Build GRCh38
rs754890926 Chr.16: 30927470-30927470 on Build GRCh38
rs781059280 Chr.16: 30927648-30927648 on Build GRCh38
rs758592580 Chr.16: 30927431-30927431 on Build GRCh38
rs1047017254 Chr.16: 30927511-30927511 on Build GRCh38
rs892718876 Chr.16: 30927481-30927481 on Build GRCh38
rs983271232 Chr.16: 30927436-30927436 on Build GRCh38
rs77477496 Chr.16: 30927473-30927473 on Build GRCh38
rs117613645 Chr.16: 30927562-30927562 on Build GRCh38
rs769601680 Chr.16: 30927474-30927474 on Build GRCh38
rs758635419 Chr.16: 30927587-30927587 on Build GRCh38
rs758980738 Chr.16: 30927501-30927501 on Build GRCh38
rs200017689 Chr.16: 30927641-30927641 on Build GRCh38
rs866602269 Chr.16: 30927619-30927619 on Build GRCh38
rs370942299 Chr.16: 30927553-30927553 on Build GRCh38
rs755082599 Chr.16: 30927647-30927647 on Build GRCh38
rs750822303 Chr.16: 30927574-30927574 on Build GRCh38
rs570318785 Chr.16: 30927565-30927565 on Build GRCh38
rs748057914 Chr.16: 30927650-30927650 on Build GRCh38
rs200698040 Chr.16: 30927608-30927608 on Build GRCh38
rs370755183 Chr.16: 30927471-30927471 on Build GRCh38
rs899205477 Chr.16: 30927605-30927605 on Build GRCh38
rs373327143 Chr.16: 30927439-30927439 on Build GRCh38
rs773897978 Chr.16: 30927493-30927493 on Build GRCh38
rs748938431 Chr.16: 30927483-30927483 on Build GRCh38
rs764626008 Chr.16: 30927529-30927529 on Build GRCh38
rs991600980 Chr.16: 30927509-30927509 on Build GRCh38
rs367643276 Chr.16: 30927594-30927594 on Build GRCh38
rs376844315 Chr.16: 30927441-30927441 on Build GRCh38

View Full Product Details