Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 157162743-157162996 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM71B
Entrez Gene ID
153745
Gene Name
family with sequence similarity 71 member B
Gene Aliases
-
UniGene
Hs.666099
Gene Chromosome Location
Chr.5: 157162332-157166268 on Build GRCh38
Transcripts
NM_130899.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
281009 c.1331C>A p.S444Y Chr.5: 157162934-157162934 on Build GRCh38
4683171 c.1380G>T p.K460N Chr.5: 157162885-157162885 on Build GRCh38
1435521 c.1458C>T p.S486S Chr.5: 157162807-157162807 on Build GRCh38
3975361 c.1474C>T p.R492* Chr.5: 157162791-157162791 on Build GRCh38
1064905 c.1485C>A p.S495S Chr.5: 157162780-157162780 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs115938677 Chr.5: 157162926-157162926 on Build GRCh38
rs779402784 Chr.5: 157162858-157162858 on Build GRCh38
rs751459650 Chr.5: 157162885-157162885 on Build GRCh38
rs372740796 Chr.5: 157162803-157162803 on Build GRCh38
rs780510911 Chr.5: 157162938-157162952 on Build GRCh38
rs775886064 Chr.5: 157162862-157162862 on Build GRCh38
rs765964858 Chr.5: 157162772-157162772 on Build GRCh38
rs375898947 Chr.5: 157162963-157162963 on Build GRCh38
rs780817844 Chr.5: 157162785-157162785 on Build GRCh38
rs756195281 Chr.5: 157162766-157162766 on Build GRCh38
rs766229012 Chr.5: 157162879-157162879 on Build GRCh38
rs758729717 Chr.5: 157162900-157162900 on Build GRCh38
rs755742422 Chr.5: 157162893-157162893 on Build GRCh38
rs752804834 Chr.5: 157162776-157162777 on Build GRCh38
rs148949971 Chr.5: 157162827-157162827 on Build GRCh38
rs765278429 Chr.5: 157162767-157162767 on Build GRCh38
rs769872026 Chr.5: 157162924-157162924 on Build GRCh38
rs140911228 Chr.5: 157162890-157162890 on Build GRCh38
rs752390382 Chr.5: 157162892-157162892 on Build GRCh38
rs868213784 Chr.5: 157162781-157162781 on Build GRCh38
rs780725075 Chr.5: 157162847-157162847 on Build GRCh38
rs372173223 Chr.5: 157162797-157162797 on Build GRCh38
rs559552263 Chr.5: 157162775-157162775 on Build GRCh38
rs529901585 Chr.5: 157162898-157162898 on Build GRCh38
rs145584571 Chr.5: 157162804-157162804 on Build GRCh38
rs201052186 Chr.5: 157162860-157162860 on Build GRCh38
rs758298028 Chr.5: 157162774-157162774 on Build GRCh38
rs759273697 Chr.5: 157162887-157162887 on Build GRCh38
rs749843966 Chr.5: 157162855-157162855 on Build GRCh38
rs374886824 Chr.5: 157162786-157162786 on Build GRCh38
rs368867239 Chr.5: 157162873-157162873 on Build GRCh38
rs778276804 Chr.5: 157162791-157162791 on Build GRCh38
rs753479388 Chr.5: 157162955-157162955 on Build GRCh38
rs375482078 Chr.5: 157162801-157162801 on Build GRCh38
rs775252520 Chr.5: 157162944-157162944 on Build GRCh38
rs768903129 Chr.5: 157162868-157162868 on Build GRCh38
rs534409939 Chr.5: 157162769-157162769 on Build GRCh38
rs746344491 Chr.5: 157162899-157162899 on Build GRCh38
rs369862230 Chr.5: 157162894-157162894 on Build GRCh38
rs748193511 Chr.5: 157162921-157162921 on Build GRCh38
rs768991205 Chr.5: 157162910-157162910 on Build GRCh38
rs762773856 Chr.5: 157162828-157162828 on Build GRCh38
rs779678953 Chr.5: 157162776-157162776 on Build GRCh38
rs201158406 Chr.5: 157162919-157162919 on Build GRCh38
rs201677910 Chr.5: 157162874-157162874 on Build GRCh38
rs777235522 Chr.5: 157162852-157162852 on Build GRCh38
rs201705403 Chr.5: 157162863-157162863 on Build GRCh38
rs754625757 Chr.5: 157162851-157162851 on Build GRCh38
rs975055330 Chr.5: 157162846-157162846 on Build GRCh38
rs747288663 Chr.5: 157162906-157162906 on Build GRCh38
rs1055104938 Chr.5: 157162949-157162949 on Build GRCh38
rs367950062 Chr.5: 157162796-157162796 on Build GRCh38
rs771331137 Chr.5: 157162857-157162857 on Build GRCh38
rs767349540 Chr.5: 157162937-157162937 on Build GRCh38
rs754569329 Chr.5: 157162783-157162783 on Build GRCh38
rs754739653 Chr.5: 157162838-157162838 on Build GRCh38
rs766250626 Chr.5: 157162831-157162831 on Build GRCh38
rs763011132 Chr.5: 157162877-157162877 on Build GRCh38
rs144873852 Chr.5: 157162925-157162925 on Build GRCh38
rs764229512 Chr.5: 157162821-157162821 on Build GRCh38
rs149717280 Chr.5: 157162798-157162798 on Build GRCh38
rs184587958 Chr.5: 157162904-157162904 on Build GRCh38
rs372413919 Chr.5: 157162950-157162950 on Build GRCh38
rs368629528 Chr.5: 157162849-157162849 on Build GRCh38
rs35758261 Chr.5: 157162837-157162837 on Build GRCh38
rs966410657 Chr.5: 157162806-157162806 on Build GRCh38
rs893412817 Chr.5: 157162968-157162968 on Build GRCh38
rs769240870 Chr.5: 157162790-157162790 on Build GRCh38
rs749935001 Chr.5: 157162768-157162768 on Build GRCh38
rs1033306473 Chr.5: 157162787-157162787 on Build GRCh38
rs561046616 Chr.5: 157162895-157162895 on Build GRCh38

View Full Product Details