Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 231398697-231398954 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
B3GNT7
Entrez Gene ID
93010
Gene Name
UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 7
Gene Aliases
beta3GnT7
UniGene
Hs.299329
Gene Chromosome Location
Chr.2: 231395624-231401164 on Build GRCh38
Transcripts
NM_145236.2 XM_005246953.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6609343 c.1015G>A p.V339M Chr.2: 231398734-231398734 on Build GRCh38
6609344 c.1061G>A p.R354Q Chr.2: 231398780-231398780 on Build GRCh38
3972077 c.982G>A p.V328I Chr.2: 231398701-231398701 on Build GRCh38
5004540 c.1025C>T p.T342M Chr.2: 231398744-231398744 on Build GRCh38
3048885 c.994A>G p.M332V Chr.2: 231398713-231398713 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749502500 Chr.2: 231398855-231398856 on Build GRCh38
rs947485523 Chr.2: 231398889-231398889 on Build GRCh38
rs754813612 Chr.2: 231398833-231398833 on Build GRCh38
rs751863553 Chr.2: 231398851-231398851 on Build GRCh38
rs376052648 Chr.2: 231398769-231398769 on Build GRCh38
rs763633234 Chr.2: 231398746-231398746 on Build GRCh38
rs777893525 Chr.2: 231398786-231398786 on Build GRCh38
rs369358770 Chr.2: 231398785-231398785 on Build GRCh38
rs748968982 Chr.2: 231398810-231398810 on Build GRCh38
rs775160836 Chr.2: 231398733-231398733 on Build GRCh38
rs781471024 Chr.2: 231398855-231398855 on Build GRCh38
rs761728771 Chr.2: 231398758-231398758 on Build GRCh38
rs578262437 Chr.2: 231398736-231398736 on Build GRCh38
rs760569261 Chr.2: 231398827-231398827 on Build GRCh38
rs560427683 Chr.2: 231398780-231398780 on Build GRCh38
rs375567551 Chr.2: 231398795-231398795 on Build GRCh38
rs1043158785 Chr.2: 231398897-231398898 on Build GRCh38
rs200686775 Chr.2: 231398906-231398906 on Build GRCh38
rs766169009 Chr.2: 231398927-231398927 on Build GRCh38
rs567694860 Chr.2: 231398907-231398907 on Build GRCh38
rs371784374 Chr.2: 231398762-231398762 on Build GRCh38
rs367672874 Chr.2: 231398745-231398745 on Build GRCh38
rs372674294 Chr.2: 231398933-231398933 on Build GRCh38
rs145105825 Chr.2: 231398820-231398820 on Build GRCh38
rs1044005987 Chr.2: 231398744-231398744 on Build GRCh38
rs757645962 Chr.2: 231398871-231398880 on Build GRCh38
rs201884921 Chr.2: 231398789-231398789 on Build GRCh38
rs767032436 Chr.2: 231398824-231398824 on Build GRCh38
rs775762806 Chr.2: 231398801-231398801 on Build GRCh38
rs1023585701 Chr.2: 231398936-231398936 on Build GRCh38
rs766135401 Chr.2: 231398770-231398770 on Build GRCh38
rs549385235 Chr.2: 231398866-231398866 on Build GRCh38
rs570729964 Chr.2: 231398825-231398825 on Build GRCh38
rs773993910 Chr.2: 231398819-231398819 on Build GRCh38
rs770086291 Chr.2: 231398731-231398731 on Build GRCh38
rs765145630 Chr.2: 231398929-231398929 on Build GRCh38
rs757418763 Chr.2: 231398794-231398794 on Build GRCh38
rs769092554 Chr.2: 231398876-231398876 on Build GRCh38
rs768254739 Chr.2: 231398811-231398811 on Build GRCh38
rs903153509 Chr.2: 231398905-231398905 on Build GRCh38
rs751138601 Chr.2: 231398749-231398749 on Build GRCh38
rs757363376 Chr.2: 231398854-231398854 on Build GRCh38
rs535210006 Chr.2: 231398916-231398916 on Build GRCh38
rs759914870 Chr.2: 231398721-231398721 on Build GRCh38
rs368886748 Chr.2: 231398928-231398928 on Build GRCh38
rs771538984 Chr.2: 231398911-231398911 on Build GRCh38
rs762169883 Chr.2: 231398934-231398934 on Build GRCh38
rs758613279 Chr.2: 231398783-231398783 on Build GRCh38
rs975426599 Chr.2: 231398862-231398862 on Build GRCh38
rs369693152 Chr.2: 231398798-231398798 on Build GRCh38
rs766191489 Chr.2: 231398832-231398832 on Build GRCh38
rs138982818 Chr.2: 231398865-231398865 on Build GRCh38
rs772774871 Chr.2: 231398913-231398913 on Build GRCh38

View Full Product Details