Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 91288948-91289195 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC88C
Entrez Gene ID
440193
Gene Name
coiled-coil domain containing 88C
Gene Aliases
DAPLE, HKRP2, KIAA1509, SCA40
UniGene
Hs.525536
Gene Chromosome Location
Chr.14: 91271323-91418498 on Build GRCh38
Transcripts
NM_001080414.3 XM_005267691.4 XM_011536796.2 XM_017021335.1 XM_017021336.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3968959 c.4417G>A p.A1473T Chr.14: 91289129-91289129 on Build GRCh38
4987903 c.4433T>C p.V1478A Chr.14: 91289113-91289113 on Build GRCh38
4571363 c.4401T>C p.C1467C Chr.14: 91289145-91289145 on Build GRCh38
3498734 c.4430C>T p.S1477F Chr.14: 91289116-91289116 on Build GRCh38
6521737 c.4417G>T p.A1473S Chr.14: 91289129-91289129 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs987768804 Chr.14: 91289014-91289014 on Build GRCh38
rs956598952 Chr.14: 91288987-91288987 on Build GRCh38
rs887378253 Chr.14: 91289058-91289058 on Build GRCh38
rs758735271 Chr.14: 91289123-91289123 on Build GRCh38
rs956532120 Chr.14: 91288988-91288988 on Build GRCh38
rs762005222 Chr.14: 91288989-91288989 on Build GRCh38
rs1004430368 Chr.14: 91289073-91289073 on Build GRCh38
rs373541390 Chr.14: 91289077-91289077 on Build GRCh38
rs766273602 Chr.14: 91289158-91289158 on Build GRCh38
rs760653768 Chr.14: 91289155-91289155 on Build GRCh38
rs142713170 Chr.14: 91289089-91289089 on Build GRCh38
rs750306032 Chr.14: 91289107-91289107 on Build GRCh38
rs575083860 Chr.14: 91288981-91288981 on Build GRCh38
rs376820989 Chr.14: 91289096-91289096 on Build GRCh38
rs914875363 Chr.14: 91289015-91289015 on Build GRCh38
rs370030976 Chr.14: 91289111-91289111 on Build GRCh38
rs771072367 Chr.14: 91289066-91289066 on Build GRCh38
rs765521764 Chr.14: 91289083-91289083 on Build GRCh38
rs757901630 Chr.14: 91289174-91289174 on Build GRCh38
rs560059428 Chr.14: 91289084-91289084 on Build GRCh38
rs377506651 Chr.14: 91289169-91289169 on Build GRCh38
rs1003395204 Chr.14: 91288982-91288982 on Build GRCh38
rs767367406 Chr.14: 91289097-91289097 on Build GRCh38
rs137977562 Chr.14: 91289046-91289046 on Build GRCh38
rs576898017 Chr.14: 91289029-91289029 on Build GRCh38
rs759883916 Chr.14: 91289078-91289078 on Build GRCh38
rs75673011 Chr.14: 91289075-91289075 on Build GRCh38
rs533658106 Chr.14: 91289041-91289041 on Build GRCh38
rs778150180 Chr.14: 91289128-91289128 on Build GRCh38
rs771160491 Chr.14: 91289130-91289130 on Build GRCh38
rs754664990 Chr.14: 91289119-91289119 on Build GRCh38
rs758725568 Chr.14: 91289168-91289168 on Build GRCh38
rs913312815 Chr.14: 91289006-91289006 on Build GRCh38
rs1018693503 Chr.14: 91289137-91289137 on Build GRCh38
rs78570354 Chr.14: 91289171-91289171 on Build GRCh38
rs765029334 Chr.14: 91289043-91289043 on Build GRCh38
rs776887120 Chr.14: 91289131-91289131 on Build GRCh38
rs564413511 Chr.14: 91289033-91289033 on Build GRCh38
rs747515656 Chr.14: 91289129-91289129 on Build GRCh38
rs753324902 Chr.14: 91289116-91289116 on Build GRCh38
rs748520551 Chr.14: 91289122-91289122 on Build GRCh38
rs200081065 Chr.14: 91289162-91289162 on Build GRCh38
rs562518256 Chr.14: 91289063-91289063 on Build GRCh38
rs143556433 Chr.14: 91289026-91289026 on Build GRCh38
rs758183068 Chr.14: 91289132-91289132 on Build GRCh38
rs753841836 Chr.14: 91289027-91289027 on Build GRCh38
rs568447378 Chr.14: 91289135-91289135 on Build GRCh38
rs531540620 Chr.14: 91289121-91289121 on Build GRCh38
rs201222692 Chr.14: 91289113-91289113 on Build GRCh38
rs767301277 Chr.14: 91289143-91289143 on Build GRCh38
rs753414934 Chr.14: 91289159-91289159 on Build GRCh38
rs201606038 Chr.14: 91289134-91289134 on Build GRCh38
rs374856540 Chr.14: 91289163-91289163 on Build GRCh38
rs537096315 Chr.14: 91289136-91289136 on Build GRCh38
rs773044678 Chr.14: 91289149-91289149 on Build GRCh38

View Full Product Details