Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 39162674-39162901 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARL5C
Entrez Gene ID
390790
Gene Name
ADP ribosylation factor like GTPase 5C
Gene Aliases
ARL12
UniGene
Hs.568007
Gene Chromosome Location
Chr.17: 39156894-39166161 on Build GRCh38
Transcripts
NM_001143968.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5426422 c.160G>A p.E54K Chr.17: 39162806-39162806 on Build GRCh38
3755444 c.116A>G p.N39S Chr.17: 39162850-39162850 on Build GRCh38
3755443 c.154G>A p.V52M Chr.17: 39162812-39162812 on Build GRCh38
5130824 c.174G>A p.P58P Chr.17: 39162792-39162792 on Build GRCh38
6292410 c.165C>A p.I55I Chr.17: 39162801-39162801 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs373678556 Chr.17: 39162804-39162804 on Build GRCh38
rs184512152 Chr.17: 39162756-39162756 on Build GRCh38
rs923835662 Chr.17: 39162742-39162742 on Build GRCh38
rs752091174 Chr.17: 39162813-39162813 on Build GRCh38
rs773173691 Chr.17: 39162696-39162696 on Build GRCh38
rs776280011 Chr.17: 39162752-39162752 on Build GRCh38
rs765912991 Chr.17: 39162760-39162760 on Build GRCh38
rs116337716 Chr.17: 39162707-39162707 on Build GRCh38
rs657672 Chr.17: 39162812-39162812 on Build GRCh38
rs760455158 Chr.17: 39162697-39162697 on Build GRCh38
rs748143509 Chr.17: 39162693-39162693 on Build GRCh38
rs61749062 Chr.17: 39162793-39162793 on Build GRCh38
rs781260429 Chr.17: 39162875-39162875 on Build GRCh38
rs657723 Chr.17: 39162850-39162850 on Build GRCh38
rs907384369 Chr.17: 39162842-39162842 on Build GRCh38
rs759356522 Chr.17: 39162699-39162705 on Build GRCh38
rs976848449 Chr.17: 39162843-39162843 on Build GRCh38
rs566016510 Chr.17: 39162801-39162801 on Build GRCh38
rs544198 Chr.17: 39162776-39162776 on Build GRCh38
rs1004669888 Chr.17: 39162835-39162835 on Build GRCh38
rs536906969 Chr.17: 39162852-39162852 on Build GRCh38
rs766464149 Chr.17: 39162807-39162807 on Build GRCh38
rs796514183 Chr.17: 39162848-39162848 on Build GRCh38
rs371774362 Chr.17: 39162722-39162722 on Build GRCh38
rs931656934 Chr.17: 39162785-39162785 on Build GRCh38
rs1036937141 Chr.17: 39162841-39162841 on Build GRCh38
rs771956426 Chr.17: 39162695-39162695 on Build GRCh38
rs781633063 Chr.17: 39162863-39162863 on Build GRCh38
rs796884035 Chr.17: 39162846-39162846 on Build GRCh38

View Full Product Details