Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 81231568-81231813 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC105371925
Entrez Gene ID
105371925
Gene Name
uncharacterized LOC105371925
Gene Aliases
-
UniGene
Hs.620720
Gene Chromosome Location
Chr.17: 81228707-81232209 on Build GRCh38
Transcripts
NR_135654.1 NM_144679.2 XM_005257066.2 XM_005257067.2 XM_006721709.3 XM_006721712.2 XM_011524355.1 XM_011524356.1 XM_011524357.1 XM_011524358.2 XM_017024202.1 XM_017024203.1 XM_017024204.1

Gene Symbol
TEPSIN
Entrez Gene ID
146705
Gene Name
TEPSIN, adaptor related protein complex 4 accessory protein
Gene Aliases
C17orf56, ENTHD2
UniGene
Hs.631761
Gene Chromosome Location
Chr.17: 81228277-81239092 on Build GRCh38
Transcripts
NR_135654.1 NM_144679.2 XM_005257066.2 XM_005257067.2 XM_006721709.3 XM_006721712.2 XM_011524355.1 XM_011524356.1 XM_011524357.1 XM_011524358.2 XM_017024202.1 XM_017024203.1 XM_017024204.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4273129 c.489G>C p.L163F Chr.17: 81231652-81231652 on Build GRCh38
4757982 c.538G>A p.E180K Chr.17: 81231603-81231603 on Build GRCh38
708673 c.773G>A p.R258H Chr.17: 81231620-81231620 on Build GRCh38
3523539 c.718C>T p.L240L Chr.17: 81231675-81231675 on Build GRCh38
7227650 c.815+10C>G p.? Chr.17: 81231568-81231568 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs140496477 Chr.17: 81231676-81231676 on Build GRCh38
rs936915015 Chr.17: 81231751-81231751 on Build GRCh38
rs781563967 Chr.17: 81231696-81231696 on Build GRCh38
rs751407535 Chr.17: 81231586-81231586 on Build GRCh38
rs759403409 Chr.17: 81231594-81231594 on Build GRCh38
rs148024725 Chr.17: 81231728-81231728 on Build GRCh38
rs551027542 Chr.17: 81231754-81231754 on Build GRCh38
rs941917809 Chr.17: 81231717-81231717 on Build GRCh38
rs571952080 Chr.17: 81231746-81231746 on Build GRCh38
rs560786388 Chr.17: 81231679-81231679 on Build GRCh38
rs1007100019 Chr.17: 81231767-81231767 on Build GRCh38
rs200620957 Chr.17: 81231701-81231701 on Build GRCh38
rs912873948 Chr.17: 81231776-81231776 on Build GRCh38
rs780934134 Chr.17: 81231606-81231606 on Build GRCh38
rs530546424 Chr.17: 81231626-81231626 on Build GRCh38
rs745832108 Chr.17: 81231716-81231716 on Build GRCh38
rs112180383 Chr.17: 81231747-81231747 on Build GRCh38
rs748406265 Chr.17: 81231700-81231700 on Build GRCh38
rs774472950 Chr.17: 81231714-81231714 on Build GRCh38
rs981393479 Chr.17: 81231792-81231792 on Build GRCh38
rs569254612 Chr.17: 81231782-81231782 on Build GRCh38
rs781439264 Chr.17: 81231625-81231625 on Build GRCh38
rs773358383 Chr.17: 81231627-81231627 on Build GRCh38
rs779217440 Chr.17: 81231735-81231735 on Build GRCh38
rs138849284 Chr.17: 81231630-81231630 on Build GRCh38
rs747438452 Chr.17: 81231620-81231620 on Build GRCh38
rs1018453050 Chr.17: 81231780-81231780 on Build GRCh38
rs767337646 Chr.17: 81231603-81231603 on Build GRCh38
rs967426772 Chr.17: 81231790-81231790 on Build GRCh38
rs61745945 Chr.17: 81231621-81231621 on Build GRCh38
rs760398750 Chr.17: 81231666-81231666 on Build GRCh38
rs138308977 Chr.17: 81231605-81231605 on Build GRCh38
rs763595844 Chr.17: 81231675-81231675 on Build GRCh38
rs780595365 Chr.17: 81231687-81231687 on Build GRCh38
rs367958395 Chr.17: 81231688-81231688 on Build GRCh38
rs759454499 Chr.17: 81231659-81231659 on Build GRCh38
rs370267135 Chr.17: 81231709-81231709 on Build GRCh38
rs953484951 Chr.17: 81231725-81231725 on Build GRCh38
rs373952381 Chr.17: 81231710-81231710 on Build GRCh38
rs1052734016 Chr.17: 81231757-81231757 on Build GRCh38
rs772086416 Chr.17: 81231723-81231723 on Build GRCh38
rs1020720889 Chr.17: 81231783-81231783 on Build GRCh38
rs541063582 Chr.17: 81231629-81231629 on Build GRCh38
rs754527638 Chr.17: 81231693-81231693 on Build GRCh38
rs546828819 Chr.17: 81231724-81231724 on Build GRCh38
rs753237513 Chr.17: 81231607-81231607 on Build GRCh38
rs151331201 Chr.17: 81231619-81231619 on Build GRCh38
rs767392535 Chr.17: 81231662-81231662 on Build GRCh38
rs149377106 Chr.17: 81231635-81231635 on Build GRCh38
rs777634905 Chr.17: 81231787-81231787 on Build GRCh38
rs760320046 Chr.17: 81231739-81231739 on Build GRCh38
rs754250062 Chr.17: 81231697-81231697 on Build GRCh38
rs925506757 Chr.17: 81231750-81231750 on Build GRCh38
rs769260298 Chr.17: 81231781-81231781 on Build GRCh38
rs897462053 Chr.17: 81231745-81231745 on Build GRCh38
rs752501204 Chr.17: 81231665-81231665 on Build GRCh38

View Full Product Details