Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 75682260-75682485 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CSPG4
Entrez Gene ID
1464
Gene Name
chondroitin sulfate proteoglycan 4
Gene Aliases
HMW-MAA, MCSP, MCSPG, MEL-CSPG, MSK16, NG2
UniGene
Hs.513044
Gene Chromosome Location
Chr.15: 75674322-75712848 on Build GRCh38
Transcripts
NM_001897.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6555094 c.4861C>A p.R1621S Chr.15: 75682382-75682382 on Build GRCh38
6840692 c.4789G>A p.V1597I Chr.15: 75682454-75682454 on Build GRCh38
965054 c.4815C>G p.L1605L Chr.15: 75682428-75682428 on Build GRCh38
6289109 c.4925C>A p.A1642D Chr.15: 75682318-75682318 on Build GRCh38
6636873 c.4863T>C p.R1621R Chr.15: 75682380-75682380 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759215027 Chr.15: 75682362-75682362 on Build GRCh38
rs756362478 Chr.15: 75682316-75682316 on Build GRCh38
rs903939713 Chr.15: 75682404-75682404 on Build GRCh38
rs757373576 Chr.15: 75682354-75682354 on Build GRCh38
rs923473968 Chr.15: 75682353-75682353 on Build GRCh38
rs746025565 Chr.15: 75682308-75682308 on Build GRCh38
rs755001075 Chr.15: 75682391-75682415 on Build GRCh38
rs548700280 Chr.15: 75682430-75682430 on Build GRCh38
rs754159916 Chr.15: 75682412-75682412 on Build GRCh38
rs748699110 Chr.15: 75682344-75682344 on Build GRCh38
rs769546688 Chr.15: 75682369-75682369 on Build GRCh38
rs145039359 Chr.15: 75682338-75682338 on Build GRCh38
rs745742249 Chr.15: 75682389-75682389 on Build GRCh38
rs767985038 Chr.15: 75682455-75682455 on Build GRCh38
rs754952359 Chr.15: 75682413-75682413 on Build GRCh38
rs200695467 Chr.15: 75682438-75682438 on Build GRCh38
rs547403556 Chr.15: 75682280-75682280 on Build GRCh38
rs779270964 Chr.15: 75682343-75682343 on Build GRCh38
rs201391275 Chr.15: 75682397-75682397 on Build GRCh38
rs771489414 Chr.15: 75682447-75682447 on Build GRCh38
rs772495667 Chr.15: 75682347-75682347 on Build GRCh38
rs767261462 Chr.15: 75682393-75682393 on Build GRCh38
rs763719075 Chr.15: 75682377-75682377 on Build GRCh38
rs142074830 Chr.15: 75682382-75682382 on Build GRCh38
rs775321944 Chr.15: 75682372-75682372 on Build GRCh38
rs143855050 Chr.15: 75682381-75682381 on Build GRCh38
rs779050438 Chr.15: 75682415-75682415 on Build GRCh38
rs749309638 Chr.15: 75682322-75682322 on Build GRCh38
rs761692206 Chr.15: 75682463-75682463 on Build GRCh38
rs144753189 Chr.15: 75682432-75682432 on Build GRCh38
rs781506272 Chr.15: 75682434-75682435 on Build GRCh38
rs945474726 Chr.15: 75682379-75682379 on Build GRCh38
rs762365316 Chr.15: 75682373-75682373 on Build GRCh38
rs570779256 Chr.15: 75682454-75682454 on Build GRCh38
rs780222155 Chr.15: 75682411-75682411 on Build GRCh38

View Full Product Details