Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 69053967-69054204 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC102B
Entrez Gene ID
79839
Gene Name
coiled-coil domain containing 102B
Gene Aliases
ACY1L, AN, C18orf14, HsT1731
UniGene
Hs.280781
Gene Chromosome Location
Chr.18: 68715221-69214350 on Build GRCh38
Transcripts
NM_001093729.1 NM_024781.2 XM_011526174.2 XM_017025973.1 XM_017025974.1 XM_017025975.1 XM_017025976.1 XM_017025977.1 XM_017025978.1 XM_017025979.1 XM_017025980.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5603088 c.1492G>A p.E498K Chr.18: 69054087-69054087 on Build GRCh38
274396 c.1496G>A p.R499K Chr.18: 69054091-69054091 on Build GRCh38
6894753 c.1501G>C p.E501Q Chr.18: 69054096-69054096 on Build GRCh38
298226 c.1537T>C p.W513R Chr.18: 69054132-69054132 on Build GRCh38
5962385 c.1459C>T p.R487C Chr.18: 69054054-69054054 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749352352 Chr.18: 69054063-69054064 on Build GRCh38
rs895112123 Chr.18: 69054054-69054054 on Build GRCh38
rs532190504 Chr.18: 69054021-69054021 on Build GRCh38
rs759297157 Chr.18: 69054066-69054066 on Build GRCh38
rs747956429 Chr.18: 69054152-69054152 on Build GRCh38
rs751886467 Chr.18: 69054074-69054074 on Build GRCh38
rs768785331 Chr.18: 69054137-69054138 on Build GRCh38
rs1019436500 Chr.18: 69054116-69054116 on Build GRCh38
rs760156587 Chr.18: 69054045-69054045 on Build GRCh38
rs745941521 Chr.18: 69054002-69054002 on Build GRCh38
rs547792221 Chr.18: 69054145-69054145 on Build GRCh38
rs781278290 Chr.18: 69054000-69054000 on Build GRCh38
rs764507471 Chr.18: 69054072-69054072 on Build GRCh38
rs749460894 Chr.18: 69054121-69054121 on Build GRCh38
rs757716714 Chr.18: 69054094-69054094 on Build GRCh38
rs774151464 Chr.18: 69054022-69054022 on Build GRCh38
rs1011626833 Chr.18: 69053996-69053996 on Build GRCh38
rs770052240 Chr.18: 69054003-69054003 on Build GRCh38
rs536746281 Chr.18: 69054009-69054009 on Build GRCh38
rs144826581 Chr.18: 69054109-69054109 on Build GRCh38
rs757730144 Chr.18: 69053999-69053999 on Build GRCh38
rs749051251 Chr.18: 69054018-69054018 on Build GRCh38
rs1033650036 Chr.18: 69054041-69054041 on Build GRCh38
rs760198313 Chr.18: 69054170-69054170 on Build GRCh38
rs977681765 Chr.18: 69054069-69054069 on Build GRCh38
rs200873817 Chr.18: 69054037-69054037 on Build GRCh38
rs771905343 Chr.18: 69054040-69054040 on Build GRCh38
rs778396838 Chr.18: 69053993-69053993 on Build GRCh38
rs768879260 Chr.18: 69054133-69054133 on Build GRCh38
rs772028652 Chr.18: 69054154-69054154 on Build GRCh38
rs1003769855 Chr.18: 69054025-69054025 on Build GRCh38
rs761751584 Chr.18: 69054038-69054038 on Build GRCh38
rs781617598 Chr.18: 69054095-69054095 on Build GRCh38
rs772772006 Chr.18: 69054042-69054042 on Build GRCh38
rs966805366 Chr.18: 69054059-69054059 on Build GRCh38
rs113120460 Chr.18: 69054014-69054014 on Build GRCh38
rs556973685 Chr.18: 69054056-69054056 on Build GRCh38
rs140831183 Chr.18: 69054105-69054105 on Build GRCh38
rs773069140 Chr.18: 69054168-69054168 on Build GRCh38
rs113651707 Chr.18: 69054055-69054055 on Build GRCh38
rs747482808 Chr.18: 69053995-69053995 on Build GRCh38

View Full Product Details