Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 14206597-14206870 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COX10
Entrez Gene ID
1352
Gene Name
COX10, heme A:farnesyltransferase cytochrome c oxidase assembly factor
Gene Aliases
-
UniGene
Hs.462278
Gene Chromosome Location
Chr.17: 14069402-14208679 on Build GRCh38
Transcripts
NM_001303.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
436056 c.966_967insT p.P324fs*12 Chr.17: 14206847-14206848 on Build GRCh38
393930 c.959C>T p.S320F Chr.17: 14206840-14206840 on Build GRCh38
7303830 c.930C>T p.G310G Chr.17: 14206811-14206811 on Build GRCh38
5021935 c.985C>T p.L329L Chr.17: 14206866-14206866 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs753721312 Chr.17: 14206767-14206767 on Build GRCh38
rs563836689 Chr.17: 14206623-14206623 on Build GRCh38
rs780244362 Chr.17: 14206661-14206661 on Build GRCh38
rs767217998 Chr.17: 14206850-14206850 on Build GRCh38
rs144296730 Chr.17: 14206798-14206799 on Build GRCh38
rs12601148 Chr.17: 14206683-14206683 on Build GRCh38
rs552570357 Chr.17: 14206663-14206663 on Build GRCh38
rs958197171 Chr.17: 14206656-14206656 on Build GRCh38
rs1056781396 Chr.17: 14206759-14206759 on Build GRCh38
rs955652435 Chr.17: 14206639-14206639 on Build GRCh38
rs184365786 Chr.17: 14206811-14206811 on Build GRCh38
rs941852186 Chr.17: 14206777-14206777 on Build GRCh38
rs756155508 Chr.17: 14206632-14206632 on Build GRCh38
rs879088846 Chr.17: 14206682-14206682 on Build GRCh38
rs535916864 Chr.17: 14206763-14206763 on Build GRCh38
rs976733487 Chr.17: 14206716-14206716 on Build GRCh38
rs765682035 Chr.17: 14206799-14206799 on Build GRCh38
rs761802668 Chr.17: 14206797-14206798 on Build GRCh38
rs971851859 Chr.17: 14206668-14206668 on Build GRCh38
rs200472593 Chr.17: 14206812-14206812 on Build GRCh38
rs76150682 Chr.17: 14206752-14206752 on Build GRCh38
rs763059631 Chr.17: 14206804-14206804 on Build GRCh38
rs374312627 Chr.17: 14206788-14206788 on Build GRCh38
rs999269753 Chr.17: 14206636-14206636 on Build GRCh38
rs1044947364 Chr.17: 14206737-14206737 on Build GRCh38
rs756441422 Chr.17: 14206803-14206804 on Build GRCh38
rs755168061 Chr.17: 14206835-14206835 on Build GRCh38
rs3785659 Chr.17: 14206647-14206647 on Build GRCh38
rs62052075 Chr.17: 14206803-14206803 on Build GRCh38
rs951437157 Chr.17: 14206738-14206738 on Build GRCh38
rs569443344 Chr.17: 14206761-14206761 on Build GRCh38
rs928053009 Chr.17: 14206675-14206675 on Build GRCh38
rs961978402 Chr.17: 14206667-14206667 on Build GRCh38
rs4792446 Chr.17: 14206724-14206724 on Build GRCh38
rs911561831 Chr.17: 14206666-14206666 on Build GRCh38
rs1018502850 Chr.17: 14206670-14206670 on Build GRCh38

View Full Product Details