Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 126504237-126504466 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C10orf90
Entrez Gene ID
118611
Gene Name
chromosome 10 open reading frame 90
Gene Aliases
FATS, bA422P15.2
UniGene
Hs.587663
Gene Chromosome Location
Chr.10: 126424997-126671038 on Build GRCh38
Transcripts
NM_001004298.2 XM_005269491.1 XM_011539216.1 XM_011539217.2 XM_011539220.2 XM_011539222.2 XM_011539223.2 XM_011539224.2 XM_011539226.1 XM_011539227.2 XM_011539230.2 XM_017015634.1 XM_017015635.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5836494 c.1139C>G p.P380R Chr.10: 126504352-126504352 on Build GRCh38
212631 c.826G>C p.E276Q Chr.10: 126504374-126504374 on Build GRCh38
4874466 c.1242A>G p.E414E Chr.10: 126504249-126504249 on Build GRCh38
6489426 c.1189A>T p.R397* Chr.10: 126504302-126504302 on Build GRCh38
4784306 c.1049A>T p.H350L Chr.10: 126504442-126504442 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs963833996 Chr.10: 126504320-126504320 on Build GRCh38
rs569578935 Chr.10: 126504413-126504413 on Build GRCh38
rs771732623 Chr.10: 126504378-126504378 on Build GRCh38
rs199768524 Chr.10: 126504279-126504279 on Build GRCh38
rs747239503 Chr.10: 126504286-126504286 on Build GRCh38
rs747764286 Chr.10: 126504336-126504336 on Build GRCh38
rs749560926 Chr.10: 126504391-126504391 on Build GRCh38
rs778895528 Chr.10: 126504343-126504343 on Build GRCh38
rs750087157 Chr.10: 126504266-126504266 on Build GRCh38
rs748116581 Chr.10: 126504354-126504354 on Build GRCh38
rs771016575 Chr.10: 126504289-126504289 on Build GRCh38
rs78267894 Chr.10: 126504384-126504384 on Build GRCh38
rs753514506 Chr.10: 126504442-126504442 on Build GRCh38
rs139034793 Chr.10: 126504257-126504257 on Build GRCh38
rs554132273 Chr.10: 126504316-126504316 on Build GRCh38
rs776638669 Chr.10: 126504292-126504292 on Build GRCh38
rs768678284 Chr.10: 126504281-126504281 on Build GRCh38
rs772739499 Chr.10: 126504421-126504421 on Build GRCh38
rs201064708 Chr.10: 126504310-126504310 on Build GRCh38
rs375882743 Chr.10: 126504277-126504277 on Build GRCh38
rs758322783 Chr.10: 126504368-126504368 on Build GRCh38
rs551343896 Chr.10: 126504298-126504298 on Build GRCh38
rs774149776 Chr.10: 126504287-126504287 on Build GRCh38
rs373996401 Chr.10: 126504303-126504303 on Build GRCh38
rs780615092 Chr.10: 126504261-126504261 on Build GRCh38
rs145104136 Chr.10: 126504327-126504327 on Build GRCh38
rs149866472 Chr.10: 126504301-126504301 on Build GRCh38
rs146720471 Chr.10: 126504423-126504423 on Build GRCh38
rs11245007 Chr.10: 126504416-126504416 on Build GRCh38
rs577186477 Chr.10: 126504326-126504327 on Build GRCh38
rs377632627 Chr.10: 126504376-126504376 on Build GRCh38
rs200126332 Chr.10: 126504340-126504340 on Build GRCh38
rs796208904 Chr.10: 126504272-126504272 on Build GRCh38
rs866371518 Chr.10: 126504371-126504371 on Build GRCh38
rs766038576 Chr.10: 126504439-126504439 on Build GRCh38
rs767041687 Chr.10: 126504335-126504335 on Build GRCh38
rs776592633 Chr.10: 126504436-126504437 on Build GRCh38
rs755780454 Chr.10: 126504274-126504274 on Build GRCh38
rs183484091 Chr.10: 126504407-126504407 on Build GRCh38
rs772826714 Chr.10: 126504401-126504401 on Build GRCh38
rs779019828 Chr.10: 126504283-126504283 on Build GRCh38

View Full Product Details