Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 104032859-104033084 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COL17A1
Entrez Gene ID
1308
Gene Name
collagen type XVII alpha 1
Gene Aliases
BA16H23.2, BP180, BPA-2, BPAG2, ERED, LAD-1
UniGene
Hs.117938
Gene Chromosome Location
Chr.10: 104031288-104085880 on Build GRCh38
Transcripts
NM_000494.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7294413 c.4296T>G p.T1432T Chr.10: 104032967-104032967 on Build GRCh38
7251357 c.4350A>G p.G1450G Chr.10: 104032913-104032913 on Build GRCh38
5836191 c.4300G>A p.G1434R Chr.10: 104032963-104032963 on Build GRCh38
3358617 c.4317C>A p.P1439P Chr.10: 104032946-104032946 on Build GRCh38
4573330 c.4317C>G p.P1439P Chr.10: 104032946-104032946 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs994356121 Chr.10: 104032884-104032884 on Build GRCh38
rs923400430 Chr.10: 104033022-104033022 on Build GRCh38
rs777927213 Chr.10: 104032928-104032928 on Build GRCh38
rs933287354 Chr.10: 104033049-104033049 on Build GRCh38
rs771186846 Chr.10: 104032934-104032934 on Build GRCh38
rs751855488 Chr.10: 104033013-104033013 on Build GRCh38
rs777690684 Chr.10: 104033011-104033011 on Build GRCh38
rs143826232 Chr.10: 104032946-104032946 on Build GRCh38
rs144093737 Chr.10: 104033001-104033001 on Build GRCh38
rs753794505 Chr.10: 104032887-104032887 on Build GRCh38
rs1050422651 Chr.10: 104033050-104033050 on Build GRCh38
rs770139408 Chr.10: 104032948-104032948 on Build GRCh38
rs778776070 Chr.10: 104032906-104032906 on Build GRCh38
rs752679841 Chr.10: 104033004-104033004 on Build GRCh38
rs759532345 Chr.10: 104032992-104032992 on Build GRCh38
rs753234705 Chr.10: 104032894-104032894 on Build GRCh38
rs758200680 Chr.10: 104032919-104032919 on Build GRCh38
rs911100679 Chr.10: 104033057-104033057 on Build GRCh38
rs772901149 Chr.10: 104032968-104032968 on Build GRCh38
rs942631190 Chr.10: 104033058-104033058 on Build GRCh38
rs747206838 Chr.10: 104032932-104032932 on Build GRCh38
rs72821434 Chr.10: 104032882-104032882 on Build GRCh38
rs77323814 Chr.10: 104033019-104033019 on Build GRCh38
rs146841330 Chr.10: 104032959-104032959 on Build GRCh38
rs55740677 Chr.10: 104032944-104032944 on Build GRCh38
rs753279573 Chr.10: 104032991-104032991 on Build GRCh38
rs796416869 Chr.10: 104033024-104033024 on Build GRCh38
rs765903384 Chr.10: 104032983-104032983 on Build GRCh38
rs758171804 Chr.10: 104033009-104033009 on Build GRCh38
rs776806814 Chr.10: 104032942-104032942 on Build GRCh38
rs201950193 Chr.10: 104032888-104032888 on Build GRCh38
rs571566750 Chr.10: 104032967-104032967 on Build GRCh38
rs1036732018 Chr.10: 104032895-104032895 on Build GRCh38
rs760269560 Chr.10: 104032971-104032971 on Build GRCh38
rs747839557 Chr.10: 104032918-104032918 on Build GRCh38
rs754832932 Chr.10: 104032896-104032896 on Build GRCh38

View Full Product Details