Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 35919875-35920116 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CFAP47
Entrez Gene ID
286464
Gene Name
cilia and flagella associated protein 47
Gene Aliases
CHDC2, CXorf22, CXorf30, CXorf59
UniGene
Hs.376425
Gene Chromosome Location
Chr.X: 35919734-36385319 on Build GRCh38
Transcripts
NM_001304548.1 NM_152632.3 XM_017029452.1 XM_017029453.1

SNP Detail

SNP ID Chromosome Location
rs757259005 Chr.X: 35920092-35920092 on Build GRCh38
rs781751927 Chr.X: 35919893-35919893 on Build GRCh38
rs767206226 Chr.X: 35919942-35919942 on Build GRCh38
rs778293818 Chr.X: 35920080-35920080 on Build GRCh38
rs143566547 Chr.X: 35919998-35919998 on Build GRCh38
rs762888297 Chr.X: 35919901-35919901 on Build GRCh38
rs756863699 Chr.X: 35919957-35919957 on Build GRCh38
rs142661727 Chr.X: 35920022-35920022 on Build GRCh38
rs763009973 Chr.X: 35919906-35919906 on Build GRCh38
rs756486257 Chr.X: 35920075-35920075 on Build GRCh38
rs761372427 Chr.X: 35920045-35920045 on Build GRCh38
rs750041803 Chr.X: 35920055-35920055 on Build GRCh38
rs771021676 Chr.X: 35920024-35920024 on Build GRCh38
rs368250675 Chr.X: 35919913-35919913 on Build GRCh38
rs188039131 Chr.X: 35920034-35920034 on Build GRCh38
rs750135715 Chr.X: 35919974-35919974 on Build GRCh38
rs6629019 Chr.X: 35919928-35919928 on Build GRCh38
rs957163235 Chr.X: 35919982-35919982 on Build GRCh38
rs771794401 Chr.X: 35920032-35920032 on Build GRCh38
rs764846948 Chr.X: 35920054-35920054 on Build GRCh38
rs774514616 Chr.X: 35920025-35920025 on Build GRCh38
rs148872967 Chr.X: 35919979-35919979 on Build GRCh38
rs758112003 Chr.X: 35920060-35920060 on Build GRCh38
rs531784634 Chr.X: 35919912-35919912 on Build GRCh38
rs906272595 Chr.X: 35919977-35919977 on Build GRCh38
rs201154448 Chr.X: 35919994-35919994 on Build GRCh38
rs752508091 Chr.X: 35919923-35919923 on Build GRCh38
rs372158027 Chr.X: 35919962-35919962 on Build GRCh38
rs753074713 Chr.X: 35920074-35920074 on Build GRCh38
rs138479255 Chr.X: 35919921-35919921 on Build GRCh38
rs755329242 Chr.X: 35919975-35919975 on Build GRCh38
rs756465850 Chr.X: 35919990-35919990 on Build GRCh38
rs1044116375 Chr.X: 35919967-35919967 on Build GRCh38
rs749825592 Chr.X: 35920087-35920087 on Build GRCh38
rs376901784 Chr.X: 35920039-35920039 on Build GRCh38
rs766014633 Chr.X: 35920070-35920070 on Build GRCh38
rs746025244 Chr.X: 35920035-35920035 on Build GRCh38
rs764057549 Chr.X: 35920036-35920036 on Build GRCh38
rs746093419 Chr.X: 35920026-35920026 on Build GRCh38
rs749366989 Chr.X: 35920023-35920023 on Build GRCh38
rs150104530 Chr.X: 35919897-35919897 on Build GRCh38
rs763993158 Chr.X: 35919931-35919931 on Build GRCh38
rs946701775 Chr.X: 35919937-35919937 on Build GRCh38

View Full Product Details